Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crybb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017268.3 | NP_001017268.1 | 3 - 4 | 289 | 68 | |
GenBank mRNA | BX703001.1 | - | 3 - 4 | 289 | 68 | |
CR855810.2 | - | 2 - 3 | 241 | 68 | ||
EL868269.1 | - | 3 - 4 | 571 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |