Room 2.16, 2nd Floor, The Frederick Douglass Centre, Newcastle University | Newcastle upon Tyne, NE4 5TG, United Kingdom
It was great seeing everyone at our Thermo Scientific Orbitrap Astral Mass Spectrometer launch event at BSPR-EuPA 2023 for lunch and networking, where we showcased how we are revolutionizing the world of proteomics and precision medicine.
The Orbitrap Astral Mass Spectrometer combines three mass analyzers: a quadrupole mass analyzer for high selectivity and high ion transmission, an Orbitrap mass analyzer for high dynamic range and high-resolution measurements, and the novel Astral analyzer for fast and sensitive measurements.
Watch our Orbitrap Astral Mass Spectrometer product video to learn more.
Lunchtime seminar
Monday, July 17 | 12:00 - 16:00 BST | Seminar room 2.16 - Lunch provided
12:00 - 13:00 | Lunch
13:00 - 16:00 | Seminar
Our topics and speakers on the 17th include:
Pushing chromatographic boundaries in proteomics using the revolutionary µ-PAC Neo columns
Speaker: Robert Van Ling, Product Manager, Thermo Fisher Scientific
A next-generation, end-to-end solution for proteomics that enables the analysis of large cohorts, powering the study of proteomics at a scale previously only obtainable by genomics
Speaker: Daniel Hermanson, Sr. Marketing Manager, Thermo Fisher Scientific
A new era in Plasma Proteomics: Introducing the Proteograph XT workflow with Orbitrap Astral MS
Speaker: Rebecca Rutherford, Director of Product Management, Seer Inc.
An advanced high-sensitivity low-nanoflow LC-MS configured for high-throughput sample-limited proteomics
Speaker: Ece Aydin, Product Specialist, Thermo Fisher Scientific
Astral building blocks
Take a quiz at our preconference seminar or visit us during the conference at Booth 10 to see the Orbitrap Astral MS in person and take part in our quiz for an additional opportunity to win the highly sought-after Orbitrap Astral MS building block kits that caused a sensation at ASMS!* Keep them for yourself or give them to someone a little younger, we won’t tell anyone.
* Required field
CMD Wide-format style fixes
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
Event date white text