Actinomycin D is an antineoplastic antibiotic which inhibits DNA-primed RNA polymerase by complexing with DNA via deoxyguanosine residues.  Actinomycin D is a widely used intercalating transcription inhibitor. This compound acts by binding to DNA at dG residues.