Search Thermo Fisher Scientific
Immunofluorescence analysis of Phospho-ATM pSer1981 was performed using 70% confluent log phase HeLa cells irradiated with UV for 4 hours. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Phospho-ATM (Ser 1981) (10H11) Mouse Monoclonal Antibody (Product ## MA12020) at 1:250 dilution in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (A28175) at a dilution of 1:2000 for 45 minutes at room temperature (Panel a: green). Nuclei (Panel b: blue) were stained with SlowFade® Gold Antifade Mountant with DAPI (Product ## S36938). F-actin (Panel c: red) was stained with Rhodamine Phalloidin (Product ## R415, 1:300). Panel d represents the merged image showing nuclear localization. Panel e shows the untreated cells. Panel f represents the no primary antibody control. The images were captured at 60X magnification.