Search Thermo Fisher Scientific
Primary Mouse Hippocampal Neurons (Cat. No. A15587) labeled with EGFP control using Lipofectamine® 2000 (Cat. No. 11668) (left), and with BacMam GFP Transduction Control (BacMam 2.0; Cat. No. B10383) (right). Cells were cultured on to poly-D-lysine coated plates for 7 days in Neurobasal® media supplemented with 2% B27 (Cat. No 17504) and 0.5 mM GLUTAMAX™-I (Cat. No. 35050).