Search
Take your work beyond the status quo with more optimization, simplicity, and assurance at every pipetting step. The Thermo Scientific FluidEase electronic pipettes are designed to help revolutionize the user experience with touchscreen interface for easy programming, ergonomic design to help reduce strain while pipetting, and accurate results with precise pipetting.
| Touchscreen | Dispensing function | Intuitive interface |
![]() |
![]() |
![]() |
2.0 Color IPS LCD screen. Capacitive touch that functions with nitrile, latex and vinyl gloves. The cover glass harness offers protection. |
7- preset dispensing functions and step by step programing. Holds up to 30 programs in the pipette. |
Icon based interface offers clear images of the function. Move from function to function with a simple tab. |
| Lightweight | Index finger operation | Ejection button |
![]() |
![]() |
![]() |
Lightweight electronic pipette that feels like holding a manual pipette. |
Operate trigger to aspirate and dispense liquid. Reduces thumb use and strain in pipetting. |
LED light illuminates the tip ejection key when tip removal is possible. Helps prevents the loss of precious samples. |
| Rotating finger rest and trigger | Rotatable lower module | Non-slip soft grip |
![]() |
![]() |
![]() |
Trigger and finger rest with broader adjustability range. Allowing left and right-handed user to pipette without strain or discomfort. |
The multichannel pipette lower module rotates so you can pipette vertically or horizontally in a plate without twisting your wrist. |
Non-slip grip for bare hands or gloves, making pipetting comfortable. |
ClipTip technology |
Control parameters
|
The interlocking technology has an interface between the pipette fitting and tips that seal by clipping into place. |
Control pipetting function such as aspirating and dispensing speed with a touch. Swipe and access the Quick Set edit screen. |
Custom calibration
|
|
| End user can create custom calibration to adjust for liquids of different density or viscosity to improve accuracy and precision. | |
| Pro ClipTip single channel |
Pro ClipTip multichannel |
Pro Finnpipette single channel |
Pro Finnpipette multichannel |
|---|---|---|---|
![]() |
![]() |
![]() |
![]() |
Volume ranges:
|
Volume ranges: 8 channel
12 channel
16 channel |
Volume ranges: 10.5-10 ul 2.5-100 ul 5-200 ul 10-300 ul 20-1000 ul 0.25-5 ml 0.5-10ml |
Volume range: 8-channel 0.5-10 ul 2.5-100 ul 5-200 ul 10-300 ul 25-1250 ul 12- channel 0.5-10 ul 2.5-100 ul 5-200 ul 10-300 ul 25-1250 ul |
| Connectivity | Modular charging rack | ClipTip pipette tips |
![]() |
![]() |
![]() |
Program your pipette from the convenience of your desk with My Pipette Creator for FluidEase. The program can be easily transferred to another pipette. |
Stand can be assembled into either 1-, 2-, 3- or 4- position stand. It also has two pipette placement position, allowing pipettes to be placed on the stand with tips attached. | ClipTip pipette tips feature “clips” that lock tips in place with a light touch for a secure seal on every channel. Tips will not loosen or fall off and will only release when ejected using minimal application pressure, making daily pipetting a totally new experience. No more banging tips on your pipette to ensure they are properly attached and sealed. |
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.