SOLA iQ Online Sulfur Analyzer
Thermo Scientific™
SOLA iQ Online Sulfur Analyzer
Determine the total sulfur content of liquid or gas phase samples with the Thermo Scientific™ SOLA iQ Online Sulfur Analyzer.
Have Questions?
Catalog NumberProduct Type
SOLAIQSOLA iQ Online Sulfur Analyzer
Catalog number SOLAIQ
Price (USD)
-
Product Type:
SOLA iQ Online Sulfur Analyzer
Determine the total sulfur concentration in liquid or gas phase samples to ensure process optimization, maximize profitability, and maintain regulatory compliance with the Thermo Scientific™ SOLA iQ Online Sulfur Analyzer. Utilizing unique Pulsed Ultraviolet Fluorescence (PUVF) technology and the proven Pyrolysis system, the SOLA iQ Analyzer builds on the success of the SOLA II range of total sulfur analyzers with an install base of thousands of units for measuring total sulfur concentration at the world's leading oil & gas companies.
The SOLA iQ Online Sulfur Analyzer replaces expensive and time consuming laboratory sampling for measuring total sulfur concentration in refinery products including gasoline, diesel, kerosene, and jet fuel as well as many process gas samples such as Natural Gas, NGL and SynGas. The SOLA iQ Analyzer offers an online adaption of ASTM D5453, D6313, D6222 & ISO 20846.
Features
  • Measurement ranges from 2ppm to100%
  • Limits of detection as low as 25ppb
  • Intuitive color touchscreen user interface
  • Pure O2 is not required, eliminating the risks associated with oxygen use in a process environment
  • Semi-continuous operation; change in sulfur concentration indicated at every injection cycle
  • Automatic density compensation for ppm sulfur wt/wt measurements
  • Easy access for maintenance and >99%uptime
  • Continuous control of UV light intensity ensures calibration is maintained over a long period of time
SOLA iQ Applications
Clean Fuels
The superior stability and precision of the SOLA iQ Analyzer enables refiners to make timely process adjustments to enhance the economic efficiency of desulfurization and fuel blending operations.
  • SOLA iQ Liquid & SOLA iQ Liquid Trace Analyzer
Flare Gas & Condensable Vapors
The highly accurate SOLA iQ Flare Analyzer features a dynamic measuring range from 10 ppm to 100% S by volume with fast high-to-low response time, enabling reliable flare stack sulfur emission reporting.
  • SOLA iQ Condensible Vapors (CV) Analyzer
  • SOLA iQ Condensible Vapors (CV) Trace Analyzer
  • SOLA iQ Flare Analyzer
Multi-Calibration/Multi-Stream
The SOLA iQ Analyzer enables multiple streams of different sulfur concentrations (i.e., batch processing, inlet/outlet of reactors, etc.) to be measured by a single analyzer.
  • SOLA iQ Liquid Analyzer
  • SOLA iQ Vapor Analyzer
  • SOLA iQ Condensible Vapors (CV) Analyzer
  • SOLA iQ Flare Analyzer
  • SOLA iQ Trace Analyzer
Specifications
Certifications/ComplianceNEC Class I, Division 2, Groups B, C & D. T2/T3 & T4 (T4 rating is application dependent)
NEC Class I, Division 1, Groups B, C & D. T2/T3 & T4 (T4 rating is application dependent)
CSA with associated “C/US Mark” Class I, Division 2, Groups B, C & D. T2/T3 & T4 (T4 rating is application dependent)
CSA with associated “C/US Mark” Class I, Division 1, Groups B, C & D. T2/T3 & T4 (T4 rating is application dependent)
ATEX Zone 1 Ex px IIC T3
ATEX Zone 2 Ex pz IIC T3 IECEx Zone 2 Ex px IIC T3
Communication DetailsAutoCONFIG™ Software: Security protected access to all analyzer setup and configuration parameters and all process and diagnostic data. Connect via a standard PC across your Local Area Network. Download 30,000 data records (process and analyzer functions), typically up to 24 hours of records (user configurable).
Current OutputsRelay & digital outputs
8x relay outputs rated 6A at 240VAC
8x solid state relay outputs rated 0.2A at 120VAC/VDC
Depth (English)18 in.
Depth (Metric)459 mm
DisplayFront panel mounted 7 in. color touchscreen user interface to access analyser functions and diagnostic data including sulfur concentrations, oven and furnace temperatures, PMT and lamp voltages, detector flow rate and more.
Electrical Requirements100/240 V, 50/60 Hz, 18 A circuit recommended, 18 A maximum during warm-up cycle, 7 to 8 A once achieving operational temperatures
Frequency50/60 Hz
Height (English)Zone 1: 56 in., Zone 2: 45 in.
Height (Metric)Zone 1: 1420 mm, Zone 2: 1130 mm
Inputs4x inputs (user configurable as 4 to 20 mA or 0-5V)
LinearityRanges ≥10 ppm: ±2% of full scale, one sample injection per minute; ±1% of full scale, two sample injections per minute
Ranges ≤10 ppm: ±2% of full scale, two sample injections per minute
Measurement RangesSOLA iQ Liquid: Full scale ranges from 5ppm to 5% - single or dual range analyzer
SOLA iQ Vapor: Full scale ranges from 5ppm to 1% - single or dual range analyzer
SOLA iQ Condensible Vapors (CV): Full scale ranges from 5ppm to 1% - single or dual range analyzer
SOLA iQ Flare: Full scale ranges from 10ppm to 100% - dual range analyzer
SOLA iQ Trace: Full scale ranges from 2ppm to 50ppm - single or dual range analyzer
No. of Process StreamsSingle or multiple (up to 4) stream control via SOLA iQ pneumatic outputs, stream selector hardware to be supplied by customer/third party
Outputs4x 4 to 20 mA outputs
PrecisionSOLA iQ Liquid & Vapor: Ranges ≥10ppm ±2% of full scale, 1 sample injection per minute; ±1% of full scale, 2 sample injections per min
SOLA iQ Liquid & Vapor: Ranges <10ppm ±2% of full scale, two sample injections per minute
SOLA iQ Flare: ±1% of full scale, two sample injections per minute
SOLA iQ Trace: ±1% of full scale, two sample injections per minute
Response TimeEvery 1 second, typically 5-6 minutes to 90% of new value (application dependent)
Serial ConnectionsRS232 Serial Port
Width (English)24 in.
Width (Metric)610 mm
Zero Grade Air3.8 to 6.9 barg, 180 to 210 L/min. (application dependent), Oil Free, -40°C (-40°F) dew point
Ambient Temperature12°C to 40°C
Detection Limits1.25% of full scale or 125 ppb (whichever is greater)
Detector TypePulsed UV Fluorescence (PUVF) with Pyrolyzer for Total Sulfur Measurement
Product TypeSOLA iQ Online Sulfur Analyzer
Voltage100/240 V
Unit SizeEach
Frequently asked questions (FAQs)
In the iQ Series Gas Analyzer instruments, which distributed measurement and control (DMC) modules are serialized?
The serialized DMCs are unique to your analyzer and its configuration. To understand which DMCs are serialized within your instrument, navigate to the “Health Check” menu via the “Settings” button on the home screen or the health check button on the status bar. From the “Health Check” menu, click on “Status and Alarms”. The analyzer's serialized DMCs can be found by clicking on the “Serial Numbers” button.
When will the iQ Series Gas Analyzer instrument platform notify me of a maintenance requirement?
The embedded preventive maintenance schedule provides a warning when an item gets within 2 months of the replacement date. At that time, the line in the schedule will turn yellow and throw a “gear icon” notification to the status bar. The line will change from yellow to red once the item gets within 1 month of the replacement date. The “gear” icon will remain in place unless some activity is done to clear it.
Can I assign a name to the iQ Series Gas Analyzer instrument?
There are a couple of ways to uniquely identify each instrument. The “instrument ID” or Modbus device ID is a user editable, numeric string. The instrument “host name” is a user editable alpha-numeric string for mobile search functionality. The “host name” can be used to access the instrument over TCP/IP but data download file names and reports will only reference the instrument serial number.
Is there a way to obtain the configuration settings for iQ Series Gas Analyzer instruments?
The iQ Series Gas Analyzer instrument configuration settings can be downloaded from the analyzer to a USB memory device.
Do alarms prioritize in the iQ Series Gas Analyzer instruments?
There is no prioritization to the assignment of alarm or maintenance notifications by the iQ Series Gas Analyzer instruments, however, the way that those events generate email notifications is different. Upon power up, the analyzer waits 2 hrs to clear any alarms prior to sending any email notifications. Post start-up sequence, an alarm needs to be active for 5 continuous mins at which time an email notification will be generated and sent. The instrument will reflect an alarm condition on the status bar at any point that the instrument meets those alarm conditions, but an email will not be generated until any single alarm condition has existed for 5 continuous mins. Maintenance notifications generated by either predictive diagnostics or the embedded preventive maintenance schedule behave in the same manner, except that the alert must be active for 24 hrs prior to an email notification being sent.