    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Isotype Controls ›
          • Mouse IgG Isotype Controls
          Invitrogen
          Mouse IgG Isotype Control
          1 Published Figure
          View all Mouse IgG Isotype Controls
          Questions & Answers
          Cite Mouse IgG Isotype Control
          Additional Information:
          {{banner.description}}
          • Published Figures (1)
          Mouse IgG Isotype Control in Control (Ctrl)
          Group 53 Created with Sketch.
          Mouse IgG Isotype Control in Control (Ctrl)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Mouse IgG Isotype Control (10400C) in Ctrl
          Lola29M/F molecular actions. a-hrobo1 promoter-luciferase reporter assays. a The robo1 genomic map (upper) and reporter schematics (lower). The genomic distance (in kb) is indicated on the top with the reference point 0, at which the robo1 genomic fragment was fused with the luciferase-coding sequence (blue box with an arrow). Below the thick line, the exon (box)-intron (thin line) organization of robo1 is ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mouse IgG Isotype Control in Control (Ctrl)
          Product Details
          10400C
          Applications
          Tested Dilution
          Flow Cytometry (Flow)
          Assay-Dependent
          Control (Ctrl)
          Assay-Dependent
          Product Specifications
          Host/Isotype
          Mouse / IgG
          Class
          Polyclonal
          Type
          Isotype Control
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          3 mg/mL
          Amount
          15 mg
          Purification
          purified
          Storage buffer
          PBS
          Contains
          0.1% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2532980
          Target Information
          The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody's species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
          View product citations for antibody 10400C on CiteAb
          Bioinformatics
          Protein Aliases: Immunoglobulin G; ImmunoglobulinG
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline