Product References
Bcl-xL mutant promotes cartilage differentiation of BMSCs by upregulating TGF-β/BMP expression levels.
Experimental and therapeutic medicine
Xiao K,Yang L,Xie W,Gao X,Huang R,Xie M
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 11-0903-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Bcl-xL mutant promotes cartilage differentiation of BMSCs by upregulating TGF-β/BMP expression levels.
Experimental and therapeutic medicine
Xiao K,Yang L,Xie W,Gao X,Huang R,Xie M
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 11-0903-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Sex-specific adipose tissue imprinting of regulatory T cells.
Nature
Vasanthakumar A,Chisanga D,Blume J,Gloury R,Britt K,Henstridge DC,Zhan Y,Torres SV,Liene S,Collins N,Cao E,Sidwell T,Li C,Spallanzani RG,Liao Y,Beavis PA,Gebhardt T,Trevaskis N,Nutt SL,Zajac JD,Davey RA,Febbraio MA,Mathis D,Shi W,Kallies A
11-0903-82 was used in Flow Cytometry to find that sex-specific differences in Treg cells from visceral adipose tissue are determined by the tissue niche in a sex-hormone-dependent manner to limit adipose tissue inflammation.
Sun Mar 01 00:00:00 EST 2020
Cytotoxicity of nonylphenol on spermatogonial stem cells via phosphatidylinositol-3-kinase/protein kinase B/mammalian target of rapamycin pathway.
World journal of stem cells
Lei JH,Yan W,Luo CH,Guo YM,Zhang YY,Wang XH,Su XJ
11-0903 was used in Flow cytometry/Cell sorting to investigate the cytotoxic effect of NP on SSCs via the phosphatidylinositol-3-kinase/protein kinase B/mammalian target of rapamycin (PI3K/AKT/mTOR) pathway.
Fri Jun 26 00:00:00 EDT 2020
Mechanisms of interleukin 4 mediated increase in efficacy of vaccines against opioid use disorders.
NPJ vaccines
Crouse B,Robinson C,Huseby Kelcher A,Laudenbach M,Abrahante JE,Pravetoni M
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 11-0903-82) in Flow Cytometry
Thu Oct 29 00:00:00 EDT 2020
Inhibition of CXCR4 inhibits the proliferation and osteogenic potential of fibroblasts from ankylosing spondylitis via the Wnt/β‑catenin pathway.
Molecular medicine reports
He C,Li D,Gao J,Li J,Liu Z,Xu W
11-0903 was used in Flow cytometry/Cell sorting to investigate the role of C-X-C chemokine receptor type 4 (CXCR4) in ossification from patients with ankylosing spondylitis.
Mon Apr 01 00:00:00 EDT 2019
Comparison between curcumin and all-trans retinoic acid in the osteogenic differentiation of mouse bone marrow mesenchymal stem cells.
Experimental and therapeutic medicine
Ahmed MF,El-Sayed AK,Chen H,Zhao R,Yusuf MS,Zuo Q,Zhang Y,Li B
11-0903 was used in Flow cytometry/Cell sorting to examine the effects of all-trans retinoic acid and curcumin on the osteogenic differentiation of mouse bone marrow mesenchymal stem cells.
Wed May 01 00:00:00 EDT 2019
Denervation-activated STAT3-IL-6 signalling in fibro-adipogenic progenitors promotes myofibres atrophy and fibrosis.
Nature cell biology
Madaro L,Passafaro M,Sala D,Etxaniz U,Lugarini F,Proietti D,Alfonsi MV,Nicoletti C,Gatto S,De Bardi M,Rojas-García R,Giordani L,Marinelli S,Pagliarini V,Sette C,Sacco A,Puri PL
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 11-0903-82) in Immunocytochemistry
Wed Aug 01 00:00:00 EDT 2018
Denervation-activated STAT3-IL-6 signalling in fibro-adipogenic progenitors promotes myofibres atrophy and fibrosis.
Nature cell biology
Madaro L,Passafaro M,Sala D,Etxaniz U,Lugarini F,Proietti D,Alfonsi MV,Nicoletti C,Gatto S,De Bardi M,Rojas-García R,Giordani L,Marinelli S,Pagliarini V,Sette C,Sacco A,Puri PL
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 11-0903-82) in Immunocytochemistry
Wed Aug 01 00:00:00 EDT 2018
A quiescent cell population replenishes mesenchymal stem cells to drive accelerated growth in mouse incisors.
Nature communications
An Z,Sabalic M,Bloomquist RF,Fowler TE,Streelman T,Sharpe PT
Thu Jan 25 00:00:00 EST 2018
Differentiation and Protective Capacity of Virus-Specific CD8+ T Cells Suggest Murine Norovirus Persistence in an Immune-Privileged Enteric Niche.
Immunity
Tomov VT,Palko O,Lau CW,Pattekar A,Sun Y,Tacheva R,Bengsch B,Manne S,Cosma GL,Eisenlohr LC,Nice TJ,Virgin HW,Wherry EJ
11-0903 was used in Flow cytometry/Cell sorting to elucidate a strategy of immune evasion by mouse norovirus via the induction of a CD8+ T cell program normally reserved for latent pathogens and persistence in an immune-privileged enteric niche.
Tue Oct 17 00:00:00 EDT 2017
Cholesterol Accumulation in Dendritic Cells Links the Inflammasome to Acquired Immunity.
Cell metabolism
Westerterp M,Gautier EL,Ganda A,Molusky MM,Wang W,Fotakis P,Wang N,Randolph GJ,D'Agati VD,Yvan-Charvet L,Tall AR
11-0903 was used in Flow cytometry to identify an essential role of DC cholesterol efflux pathways in maintaining immune tolerance.
Tue Jun 06 00:00:00 EDT 2017
Cardiac Tissue Injury and Remodeling Is Dependent Upon MR Regulation of Activation Pathways in Cardiac Tissue Macrophages.
Endocrinology
Shen JZ,Morgan J,Tesch GH,Rickard AJ,Chrissobolis S,Drummond GR,Fuller PJ,Young MJ
11-0903 was used in Flow cytometry/Cell sorting to determine the cellular mechanisms of macrophage mineralocorticoid receptor signaling in isolated tissue macrophages.
Mon Aug 01 00:00:00 EDT 2016
Transient Expression of WNT2 Promotes Somatic Cell Reprogramming by Inducing β-Catenin Nuclear Accumulation.
Stem cell reports
Kimura M,Nakajima-Koyama M,Lee J,Nishida E
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 11-0903-82) in Flow Cytometry
Tue Jun 14 00:00:00 EDT 2016
Skin CD4(+) memory T cells exhibit combined cluster-mediated retention and equilibration with the circulation.
Nature communications
Collins N,Jiang X,Zaid A,Macleod BL,Li J,Park CO,Haque A,Bedoui S,Heath WR,Mueller SN,Kupper TS,Gebhardt T,Carbone FR
11-0903 was used in Flow cytometry/Cell sorting to asses the pattern of tissue-retention and equilibration of CD4(+) memory T cells in skin and how this is altered by infection and inflammation.
Tue May 10 00:00:00 EDT 2016
Clec11a/osteolectin is an osteogenic growth factor that promotes the maintenance of the adult skeleton.
eLife
Yue R,Shen B,Morrison SJ
11-0903 was used in Flow cytometry/Cell sorting to study how Clec11a/Osteolectin maintains the adult skeleton by promoting the differentiation of mesenchymal progenitors into mature osteoblasts.
Tue Dec 13 00:00:00 EST 2016
Gastric LTi cells promote lymphoid follicle formation but are limited by IRAK-M and do not alter microbial growth.
Mucosal immunology
Shiu J,Piazuelo MB,Ding H,Czinn SJ,Drakes ML,Banerjee A,Basappa N,Kobayashi KS,Fricke WF,Blanchard TG
11-0903 was used in Flow cytometry/Cell sorting to investigate the role of lymphoid tissue inducer cells in the gastric mucosa in the context of microbial infection.
Tue Sep 01 00:00:00 EDT 2015
Collaborative rewiring of the pluripotency network by chromatin and signalling modulating pathways.
Nature communications
Tran KA,Jackson SA,Olufs ZP,Zaidan NZ,Leng N,Kendziorski C,Roy S,Sridharan R
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 11-0903-82) in Flow Cytometry
Wed Feb 04 00:00:00 EST 2015
Intronic regulation of Aire expression by Jmjd6 for self-tolerance induction in the thymus.
Nature communications
Yanagihara T,Sanematsu F,Sato T,Uruno T,Duan X,Tomino T,Harada Y,Watanabe M,Wang Y,Tanaka Y,Nakanishi Y,Suyama M,Yoshinori F
11-0903 was used in Flow cytometry/Cell sorting to study the regulation of Aire expression in medullary thymic epithelial cells.
Wed Nov 04 00:00:00 EST 2015
Intrahepatic innate lymphoid cells secrete IL-17A and IL-17F that are crucial for T cell priming in viral infection.
Journal of immunology (Baltimore, Md. : 1950)
Jie Z,Liang Y,Hou L,Dong C,Iwakura Y,Soong L,Cong Y,Sun J
11-0903 was used in Flow cytometry/Cell sorting to investigate the source and regulation of the IL-17 surge in the liver microenvironment that occurs early in the response to viral infection.
Tue Apr 01 00:00:00 EDT 2014
Ionizing radiation-induced expression of INK4a/ARF in murine bone marrow-derived stromal cell populations interferes with bone marrow homeostasis.
Blood
Carbonneau CL,Despars G,Rojas-Sutterlin S,Fortin A,Le O,Hoang T,Beauséjour CM
11-0903 was used in Flow cytometry/Cell sorting to investigate the effect of ionizing radiation on murine bone marrow-derived stromal cell populations, and overall bone marrow homeostasis.
Thu Jan 19 00:00:00 EST 2012
Dendritic cell-specific ablation of the protein tyrosine phosphatase Shp1 promotes Th1 cell differentiation and induces autoimmunity.
Journal of immunology (Baltimore, Md. : 1950)
Kaneko T,Saito Y,Kotani T,Okazawa H,Iwamura H,Sato-Hashimoto M,Kanazawa Y,Takahashi S,Hiromura K,Kusakari S,Kaneko Y,Murata Y,Ohnishi H,Nojima Y,Takagishi K,Matozaki T
11-0903 was used in Immunohistochemistry on frozen tissues to investigate the molecules that regulate dendritic cells to protect against autoimmunity, showing that loss of Shp1 promotes Th1 cell differentiation and induces autoimmunity.
Fri Jun 01 00:00:00 EDT 2012
Delayed, asynchronous, and reversible T-lineage specification induced by Notch/Delta signaling.
Genes & development
Taghon TN,David ES,Zúñiga-Pflücker JC,Rothenberg EV
11-0903 was used in Flow cytometry/Cell sorting to study how Notch/Delta signaling is not sufficient for T-lineage specification and commitment, but can be permissive for the maintenance and proliferation of uncommitted progenitors.
Fri Apr 15 00:00:00 EDT 2005