Product References
Profiling of Tregs across tissues reveals plasticity in ST2 expression and hierarchies in tissue-specific phenotypes.
iScience
Spath S,Roan F,Presnell SR,Höllbacher B,Ziegler SF
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Fri Sep 16 00:00:00 EDT 2022
An intranasal lentiviral booster reinforces the waning mRNA vaccine-induced SARS-CoV-2 immunity that it targets to lung mucosa.
Molecular therapy : the journal of the American Society of Gene Therapy
Vesin B,Lopez J,Noirat A,Authié P,Fert I,Le Chevalier F,Moncoq F,Nemirov K,Blanc C,Planchais C,Mouquet H,Guinet F,Hardy D,Vives FL,Gerke C,Anna F,Bourgine M,Majlessi L,Charneau P
11-1031-82 was used in Flow cytometry/Cell sorting to demonstrate the suitability of the LV::SBeta-2P vaccine candidate as an intranasal booster against COVID-19. LV::SBeta-2P vaccination was also fully protective against Omicron infection of the lungs and central nervous system, in the highly susceptible B6.K18-hACE2IP-THV transgenic mice.
Wed Sep 07 00:00:00 EDT 2022
STING Activated Tumor-Intrinsic Type I Interferon Signaling Promotes CXCR3 Dependent Antitumor Immunity in Pancreatic Cancer.
Cellular and molecular gastroenterology and hepatology
Vonderhaar EP,Barnekow NS,McAllister D,McOlash L,Eid MA,Riese MJ,Tarakanova VL,Johnson BD,Dwinell MB
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Mon Jan 24 00:00:00 EST 2022
Macrophages Maintain Epithelium Integrity by Limiting Fungal Product Absorption.
Cell
Chikina AS,Nadalin F,Maurin M,San-Roman M,Thomas-Bonafos T,Li XV,Lameiras S,Baulande S,Henri S,Malissen B,Lacerda Mariano L,Barbazan J,Blander JM,Iliev ID,Matic Vignjevic D,Lennon-Duménil AM
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Thu Oct 15 00:00:00 EDT 2020
Contact-dependent delivery of IL-2 by dendritic cells to CD4 T cells in the contraction phase promotes their long-term survival.
Protein & cell
Tong D,Zhang L,Ning F,Xu Y,Hu X,Shi Y
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
Inhibition of the Wnt/β-catenin pathway enhances antitumor immunity in ovarian cancer.
Therapeutic advances in medical oncology
Doo DW,Meza-Perez S,Londoño AI,Goldsberry WN,Katre AA,Boone JD,Moore DJ,Hudson CT,Betella I,McCaw TR,Gangrade A,Bao R,Luke JJ,Yang ES,Birrer MJ,Starenki D,Cooper SJ,Buchsbaum DJ,Norian LA,Randall TD,Arend RC
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Thu Apr 14 00:00:00 EDT 2022
Human and Mouse Transcriptome Profiling Identifies Cross-Species Homology in Pulmonary and Lymph Node Mononuclear Phagocytes.
Cell reports
Leach SM,Gibbings SL,Tewari AD,Atif SM,Vestal B,Danhorn T,Janssen WJ,Wager TD,Jakubzick CV
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Tue Nov 03 00:00:00 EST 2020
Programming Multifaceted Pulmonary T Cell Immunity by Combination Adjuvants.
Cell reports. Medicine
Marinaik CB,Kingstad-Bakke B,Lee W,Hatta M,Sonsalla M,Larsen A,Neldner B,Gasper DJ,Kedl RM,Kawaoka Y,Suresh M
11-1031 was used in Flow cytometry/Cell sorting to provide further insights into vaccine-induced multifaceted mucosal T cell immunity with implications in the development of vaccines against respiratorypathogens, including influenza virus and SARS-CoV-2.
Tue Sep 22 00:00:00 EDT 2020
Acid ceramidase of macrophages traps herpes simplex virus in multivesicular bodies and protects from severe disease.
Nature communications
Lang J,Bohn P,Bhat H,Jastrow H,Walkenfort B,Cansiz F,Fink J,Bauer M,Olszewski D,Ramos-Nascimento A,Duhan V,Friedrich SK,Becker KA,Krawczyk A,Edwards MJ,Burchert A,Huber M,Friebus-Kardash J,Göthert JR,Hardt C,Probst HC,Schumacher F,Köhrer K,Kleuser B,Babiychuk EB,Sodeik B,Seibel J,Greber UF,Lang PA,Gulbins E,Lang KS
11-1031-82 was used in Flow Cytometry to show that macrophages take up HSV-1 via endocytosis and transport the virions into multivesicular bodies (MVBs).
Thu Mar 12 00:00:00 EDT 2020
Bilateral Effect of the Unilateral Corneal Nerve Cut on Both Ocular Surface and Lacrimal Gland.
Investigative ophthalmology & visual science
Lee HK,Kim KW,Ryu JS,Jeong HJ,Lee SM,Kim MK
11-1031 was used in Flow cytometry/Cell sorting to demonstrate that a unilateral cut of the corneal nerve results in activation of immune cells on the ocular surface and dysregulation of lacrimal secretion bilaterally through the bidirectional neuronal signals.
Wed Jan 02 00:00:00 EST 2019
CSF1R antagonism limits local restimulation of antiviral CD8+ T cells during viral encephalitis.
Journal of neuroinflammation
Funk KE,Klein RS
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Thu Jan 31 00:00:00 EST 2019
Virus-specific memory T cells populate tumors and can be repurposed for tumor immunotherapy.
Nature communications
Rosato PC,Wijeyesinghe S,Stolley JM,Nelson CE,Davis RL,Manlove LS,Pennell CA,Blazar BR,Chen CC,Geller MA,Vezys V,Masopust D
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Mon Feb 04 00:00:00 EST 2019
Mrp1 is involved in lipid presentation and iNKT cell activation by Streptococcus pneumoniae.
Nature communications
Chandra S,Gray J,Kiosses WB,Khurana A,Hitomi K,Crosby CM,Chawla A,Fu Z,Zhao M,Veerapen N,Richardson SK,Porcelli SA,Besra G,Howell AR,Sharma S,Peters B,Kronenberg M
11-1031 was used in Immunocytochemistry-immunoflourescence to elucidate the CD1d presentation pathway.
Mon Oct 15 00:00:00 EDT 2018
Reactive glia promote development of CD103+ CD69+ CD8+ T-cells through programmed cell death-ligand 1 (PD-L1).
Immunity, inflammation and disease
Prasad S,Hu S,Sheng WS,Chauhan P,Lokensgard JR
11-1031 was used in Flow cytometry/Cell sorting to demonstrate a role for activated glia in promoting development of bTRM through the PD-1: PD-L1 pathway.
Fri Jun 01 00:00:00 EDT 2018
NLRP12 attenuates colon inflammation by maintaining colonic microbial diversity and promoting protective commensal bacterial growth.
Nature immunology
Chen L,Wilson JE,Koenigsknecht MJ,Chou WC,Montgomery SA,Truax AD,Brickey WJ,Packey CD,Maharshak N,Matsushima GK,Plevy SE,Young VB,Sartor RB,Ting JP
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
Deletion of PIKfyve alters alveolar macrophage populations and exacerbates allergic inflammation in mice.
The EMBO journal
Kawasaki T,Ito K,Miyata H,Akira S,Kawai T
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Wed Jun 14 00:00:00 EDT 2017
Mesenteric lymph node CD11b- CD103+ PD-L1High dendritic cells highly induce regulatory T cells.
Immunology
Shiokawa A,Kotaki R,Takano T,Nakajima-Adachi H,Hachimura S
11-1031 was used in Flow cytometry/Cell sorting to investigate the expression of these molecules on MLN CD11c+ cells.
Fri Sep 01 00:00:00 EDT 2017
Precision-cut Mouse Lung Slices to Visualize Live Pulmonary Dendritic Cells.
Journal of visualized experiments : JoVE
Lyons-Cohen MR,Thomas SY,Cook DN,Nakano H
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Immunofluorescence
Wed Apr 05 00:00:00 EDT 2017
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Thu Oct 06 00:00:00 EDT 2016
Brain-resident memory T cells represent an autonomous cytotoxic barrier to viral infection.
The Journal of experimental medicine
Steinbach K,Vincenti I,Kreutzfeldt M,Page N,Muschaweckh A,Wagner I,Drexler I,Pinschewer D,Korn T,Merkler D
11-1031 was used in Immunofluorescence on frozen tissues to characterise the functioning of brain-resident memory T cells in an animal model of viral infection.
Mon Jul 25 00:00:00 EDT 2016
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2016
Diet-dependent, microbiota-independent regulation of IL-10-producing lamina propria macrophages in the small intestine.
Scientific reports
Ochi T,Feng Y,Kitamoto S,Nagao-Kitamoto H,Kuffa P,Atarashi K,Honda K,Teitelbaum DH,Kamada N
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Wed Jun 15 00:00:00 EDT 2016
A reproducible method for the expansion of mouse CD8+ T lymphocytes.
Journal of immunological methods
Lewis MD,de Leenheer E,Fishman S,Siew LK,Gross G,Wong FS
11-1031 was used in Flow cytometry/Cell sorting to report a tissue culture protocol for the reliable expansion of CD8+ T-cells derived from murine spleens.
Sun Feb 01 00:00:00 EST 2015
Immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Nature medicine
Clatworthy MR,Aronin CE,Mathews RJ,Morgan NY,Smith KG,Germain RN
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 11-1031-82) in Flow Cytometry
Mon Dec 01 00:00:00 EST 2014
Lung dendritic cells undergo maturation and polarization towards a T helper type 2-stimulating phenotype in a mouse model of asthma: Role of nerve growth factor.
Experimental and therapeutic medicine
Qin Q,Wang Z,Pan P,Cao Z,Xia Q,Tan H,Hu C
Sat Nov 01 00:00:00 EDT 2014
Systemic sensitization with the protein allergen ovalbumin augments local sensitization in atopic dermatitis.
Journal of inflammation research
Yoo J,Manicone AM,McGuire JK,Wang Y,Parks WC
11-1031 was used in Flow cytometry/Cell sorting to reveal a mouse model of atopic dermatitis that reveals novel insights into the pathogenesis of this complex disease.
Thu Mar 27 00:00:00 EDT 2014
MHC class I-restricted myelin epitopes are cross-presented by Tip-DCs that promote determinant spreading to CD8⁺ T cells.
Nature immunology
Ji Q,Castelli L,Goverman JM
11-1031 was used in Flow cytometry/Cell sorting to investigate the mechanisms underlying the role of CD8(+) T cells in the pathogenesis of multiple sclerosis.
Fri Mar 01 00:00:00 EST 2013
Protection of islet grafts through transforming growth factor-β-induced tolerogenic dendritic cells.
Diabetes
Thomas DC,Wong FS,Zaccone P,Green EA,Wållberg M
11-1031 was used in Flow cytometry/Cell sorting to investigate methods to achieve long-term β-islet graft tolerance for diabetes type 1, showing that islets can be protected through expression of TGF-β.
Sun Sep 01 00:00:00 EDT 2013
PD-L2 modulates asthma severity by directly decreasing dendritic cell IL-12 production.
Mucosal immunology
Lewkowich IP,Lajoie S,Stoffers SL,Suzuki Y,Richgels PK,Dienger K,Sproles AA,Yagita H,Hamid Q,Wills-Karp M
11-1031 was used in Flow cytometry/Cell sorting to investigate the mechanism by which PD-L2 modulates asthma severity in human and mouse lung samples.
Mon Jul 01 00:00:00 EDT 2013
Changes in lamina propria dendritic cells on the oral administration of exogenous protein antigens during weaning.
Cytotechnology
Ohue R,Nakamoto M,Kitabatake N,Tani F
11-1031 was used in Flow cytometry/Cell sorting to suggest that the behavior of tolerogenic PDCA-1(+) DCs may change at weaning with the removal of the immunoprotective components of maternal milk.
Tue May 01 00:00:00 EDT 2012
H2 control of natural T regulatory cell frequency in the lymph node correlates with susceptibility to day 3 thymectomy-induced autoimmune disease.
Journal of immunology (Baltimore, Md. : 1950)
del Rio R,Sun Y,Alard P,Tung KS,Teuscher C
11-1031 was used in Flow cytometry/Cell sorting to confirm that quantitative trait loci on a range of chromosomes control susceptibility to autoimmune ovarian dysgenesis.
Sat Jan 01 00:00:00 EST 2011
Down-regulation of Gfi-1 expression by TGF-beta is important for differentiation of Th17 and CD103+ inducible regulatory T cells.
The Journal of experimental medicine
Zhu J,Davidson TS,Wei G,Jankovic D,Cui K,Schones DE,Guo L,Zhao K,Shevach EM,Paul WE
11-1031 was used in Flow cytometry/Cell sorting to show that Gfi-1, a transcriptional repressor, has a critical role in enhancing Th2 cell expansion and repressing induction of Th17 and CD103(+) iTreg cells.
Mon Feb 16 00:00:00 EST 2009
Trafficking of antigen-specific CD8+ T lymphocytes to mucosal surfaces following intramuscular vaccination.
Journal of immunology (Baltimore, Md. : 1950)
Kaufman DR,Liu J,Carville A,Mansfield KG,Havenga MJ,Goudsmit J,Barouch DH
11-1031 was used in Flow cytometry/Cell sorting to evaluate mucosal cellular immune responses in mice and rhesus monkeys after systemic immunisation with various replication-incompetent recombinant adenovirus vectors.
Mon Sep 15 00:00:00 EDT 2008
Localization of intestinal intraepithelial T lymphocytes involves regulation of alphaEbeta7 expression by transforming growth factor-beta.
International immunology
Suzuki R,Nakao A,Kanamaru Y,Okumura K,Ogawa H,Ra C
Mon Apr 01 00:00:00 EST 2002