Product References
CD8+ T cell-Dependent Remodeling of the Tumor Microenvironment Overcomes Chemoresistance.
Cancer immunology research
Lao L,Zeng W,Huang P,Chen H,Jia Z,Wang P,Huang D,Chen J,Nie Y,Yang L,Wu W,Liu J
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Rapamycin improves Graves' orbitopathy by suppressing CD4+ cytotoxic T lymphocytes.
JCI insight
Zhang M,Chong KK,Chen ZY,Guo H,Liu YF,Kang YY,Li YJ,Shi TT,Lai KK,He MQ,Ye K,Kahaly GJ,Shi BY,Wang Y
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Flow Cytometry
Wed Feb 08 00:00:00 EST 2023
Neoadjuvant chemotherapy plus nivolumab with or without ipilimumab in operable non-small cell lung cancer: the phase 2 platform NEOSTAR trial.
Nature medicine
Cascone T,Leung CH,Weissferdt A,Pataer A,Carter BW,Godoy MCB,Feldman H,William WN,Xi Y,Basu S,Sun JJ,Yadav SS,Rojas Alvarez FR,Lee Y,Mishra AK,Chen L,Pradhan M,Guo H,Sinjab A,Zhou N,Negrao MV,Le X,Gay CM,Tsao AS,Byers LA,Altan M,Glisson BS,Fossella FV,Elamin YY,Blumenschein G,Zhang J,Skoulidis F,Wu J,Mehran RJ,Rice DC,Walsh GL,Hofstetter WL,Rajaram R,Antonoff MB,Fujimoto J,Solis LM,Parra ER,Haymaker C,Wistuba II,Swisher SG,Vaporciyan AA,Lin HY,Wang J,Gibbons DL,Jack Lee J,Ajami NJ,Wargo JA,Allison JP,Sharma P,Kadara H,Heymach JV,Sepesi B
11-9994-42 was used in Flow Cytometry, Immunohistochemistry to report the results and correlates of two arms of the phase 2 platform NEOSTAR trial testing neoadjuvant Nivo+CT and Ipi+Nivo+CT with major pathologic response (MPR) as the primary endpoint.
Wed Mar 01 00:00:00 EST 2023
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
11-9994-42 was used in Flow Cytometry to demonstrate that LINC01123 acts as a miR-214-3p sponge to inhibit the activation of CD8+T cells and promote the progression of HNSCC by upregulating B7-H3.
Thu Feb 03 00:00:00 EST 2022
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
11-9994-42 was used in Flow Cytometry to demonstrate that LINC01123 acts as a miR-214-3p sponge to inhibit the activation of CD8+T cells and promote the progression of HNSCC by upregulating B7-H3.
Thu Feb 03 00:00:00 EST 2022
Single-cell transcriptomics links malignant T cells to the tumor immune landscape in cutaneous T cell lymphoma.
Nature communications
Liu X,Jin S,Hu S,Li R,Pan H,Liu Y,Lai P,Xu D,Sun J,Liu Z,Gao Y,Zhao Y,Liu F,Xiao Y,Li Y,Wen Y,Chen Z,Xu B,Lin Y,Ran M,Li Q,Yang S,Li H,Tu P,Haniffa M,Teichmann SA,Bai F,Wang Y
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Immunohistochemistry
Thu Mar 03 00:00:00 EST 2022
PD-1-Expressing SARS-CoV-2-Specific CD8+ T Cells Are Not Exhausted, but Functional in Patients with COVID-19.
Immunity
Rha MS,Jeong HW,Ko JH,Choi SJ,Seo IH,Lee JS,Sa M,Kim AR,Joo EJ,Ahn JY,Kim JH,Song KH,Kim ES,Oh DH,Ahn MY,Choi HK,Jeon JH,Choi JP,Kim HB,Kim YK,Park SH,Choi WS,Choi JY,Peck KR,Shin EC
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Human Thymic CD10+ PD-1+ Intraepithelial Lymphocyte Precursors Acquire Interleukin-15 Responsiveness at the CD1a- CD95+ CD28- CCR7- Developmental Stage.
International journal of molecular sciences
Billiet L,Goetgeluk G,Bonte S,De Munter S,De Cock L,Pille M,Ingels J,Jansen H,Weening K,Van Nieuwerburgh F,Kerre T,Taghon T,Leclercq G,Vandekerckhove B
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Flow Cytometry
Fri Nov 20 00:00:00 EST 2020
Neutrophil expansion defines an immunoinhibitory peripheral and intratumoral inflammatory milieu in resected non-small cell lung cancer: a descriptive analysis of a prospectively immunoprofiled cohort.
Journal for immunotherapy of cancer
Mitchell KG,Diao L,Karpinets T,Negrao MV,Tran HT,Parra ER,Corsini EM,Reuben A,Federico L,Bernatchez C,Dejima H,Francisco-Cruz A,Wang J,Antonoff MB,Vaporciyan AA,Swisher SG,Cascone T,Wistuba II,Heymach JV,Gibbons DL,Zhang J,Haymaker CL,Sepesi B
11-9994 was used in Flow cytometry/Cell sorting to demonstrate that neutrophil expansion reflects protumorigenic and immunosuppressive processes that manifest as worse overall survival (OS) in patients undergoing non-small lung cancer (NSCLC) resection.
Wed Apr 01 00:00:00 EDT 2020
Enhanced Anti-lymphoma Activity of CAR19-iNKT Cells Underpinned by Dual CD19 and CD1d Targeting.
Cancer cell
Rotolo A,Caputo VS,Holubova M,Baxan N,Dubois O,Chaudhry MS,Xiao X,Goudevenou K,Pitcher DS,Petevi K,Kachramanoglou C,Iles S,Naresh K,Maher J,Karadimitris A
11-9994 was used in Flow cytometry/Cell sorting to investigate whether chimeric antigen receptor anti-CD19-engineering of the CD1d-restricted invariant natural killer T cells would result in enhanced anti-lymphoma activity.
Mon Oct 08 00:00:00 EDT 2018
Blocking the recruitment of naive CD4+ T cells reverses immunosuppression in breast cancer.
Cell research
Su S,Liao J,Liu J,Huang D,He C,Chen F,Yang L,Wu W,Chen J,Lin L,Zeng Y,Ouyang N,Cui X,Yao H,Su F,Huang JD,Lieberman J,Liu Q,Song E
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
The Transcription Factor Hobit Identifies Human Cytotoxic CD4+ T Cells.
Frontiers in immunology
Oja AE,Vieira Braga FA,Remmerswaal EB,Kragten NA,Hertoghs KM,Zuo J,Moss PA,van Lier RA,van Gisbergen KP,Hombrink P
11-9994 was used in Flow cytometry/Cell sorting to suggest a shared differentiation pathway in CD4+, CD8+, and γδ+ T cells that may involve Hobit-driven acquisition of long-lived cytotoxic effector function.
Mon Jan 31 00:00:00 EST 2022
Human Asymptomatic Epitopes Identified from the Herpes Simplex Virus Tegument Protein VP13/14 (UL47) Preferentially Recall Polyfunctional Effector Memory CD44high CD62Llow CD8+ TEM Cells and Protect Humanized HLA-A*02:01 Transgenic Mice against Ocular Herpesvirus Infection.
Journal of virology
Srivastava R,Khan AA,Garg S,Syed SA,Furness JN,Vahed H,Pham T,Yu HT,Nesburn AB,BenMohamed L
11-9994 was used in Flow cytometry/Cell sorting to outline the phenotypic and functional features of protective HSV-specific CD8 T cells that should guide the development of a safe and effective T-cell-based herpes simplex vaccine.
Sun Jan 15 00:00:00 EST 2017
Eomesodermin and T-bet mark developmentally distinct human natural killer cells.
JCI insight
Collins A,Rothman N,Liu K,Reiner SL
11-9994 was used in Flow cytometry/Cell sorting to demonstrate the expression pattern of signature NK cell transcription factors during developmental maturation.
Thu Mar 09 00:00:00 EST 2017
Peritoneal carcinomatosis of colorectal cancer is characterized by structural and functional reorganization of the tumor microenvironment inducing senescence and proliferation arrest in cancer cells.
Oncoimmunology
Seebauer CT,Brunner S,Glockzin G,Piso P,Ruemmele P,Schlitt HJ,Geissler EK,Fichtner-Feigl S,Kesselring R
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Regulatory T Cell Modulation by CBP/EP300 Bromodomain Inhibition.
The Journal of biological chemistry
Ghosh S,Taylor A,Chin M,Huang HR,Conery AR,Mertz JA,Salmeron A,Dakle PJ,Mele D,Cote A,Jayaram H,Setser JW,Poy F,Hatzivassiliou G,DeAlmeida-Nagata D,Sandy P,Hatton C,Romero FA,Chiang E,Reimer T,Crawford T,Pardo E,Watson VG,Tsui V,Cochran AG,Zawadzke L,Harmange JC,Audia JE,Bryant BM,Cummings RT,Magnuson SR,Grogan JL,Bellon SF,Albrecht BK,Sims RJ,Lora JM
Fri Jun 17 00:00:00 EDT 2016
IL12-mediated sensitizing of T-cell receptor-dependent and -independent tumor cell killing.
Oncoimmunology
Braun M,Ress ML,Yoo YE,Scholz CJ,Eyrich M,Schlegel PG,Wölfl M
11-9994 was used in Flow cytometry/Cell sorting to investigate how IL12, and other inflammatory cytokines, shape effector functions of human T-cells.
Fri Jul 01 00:00:00 EDT 2016
Cutting edge: identification and characterization of human intrahepatic CD49a+ NK cells.
Journal of immunology (Baltimore, Md. : 1950)
Marquardt N,Béziat V,Nyström S,Hengst J,Ivarsson MA,Kekäläinen E,Johansson H,Mjösberg J,Westgren M,Lankisch TO,Wedemeyer H,Ellis EC,Ljunggren HG,Michaëlsson J,Björkström NK
11-9994 was used in Flow cytometry/Cell sorting to study the unique lineage of CD49a(+)DX5(-) NK cells, showing that they are readily detectable in the liver but not in hepatic venous or peripheral blood.
Sun Mar 15 00:00:00 EDT 2015
Differentiation and functional regulation of human fetal NK cells.
The Journal of clinical investigation
Ivarsson MA,Loh L,Marquardt N,Kekäläinen E,Berglin L,Björkström NK,Westgren M,Nixon DF,Michaëlsson J
11-9994 was used in Flow cytometry/Cell sorting to provide a basis for understanding the role of foetal NK cells in pregnancy complications.
Sun Sep 01 00:00:00 EDT 2013
Lipoxin A4 regulates natural killer cell and type 2 innate lymphoid cell activation in asthma.
Science translational medicine
Barnig C,Cernadas M,Dutile S,Liu X,Perrella MA,Kazani S,Wechsler ME,Israel E,Levy BD
Published figure using Perforin monoclonal antibody (Product # 11-9994-42) in Immunofluorescence
Wed Feb 27 00:00:00 EST 2013
Transfer of regulatory properties from tolerogenic to proinflammatory dendritic cells via induced autoreactive regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Kleijwegt FS,Laban S,Duinkerken G,Joosten AM,Koeleman BP,Nikolic T,Roep BO
11-9994 was used in Flow cytometry/Cell sorting to study the mechanism by which infectious tolerance applies to human dendritic cells.
Thu Dec 15 00:00:00 EST 2011
Common gamma chain cytokines promote rapid in vitro expansion of allo-specific human CD8+ suppressor T cells.
PloS one
Yu Y,Zitzner JR,Houlihan J,Herrera N,Xu L,Miller J,Mathew JM,Tambur AR,Luo X
11-9994 was used in Flow cytometry/Cell sorting to demonstrate the use of common γc cytokines in combination with donor APCs as a strategy for generation of donor-specific T cells for immunotherapy.
Mon Aug 27 00:00:00 EDT 2012
The lytic potential of human liver NK cells is restricted by their limited expression of inhibitory killer Ig-like receptors.
Journal of immunology (Baltimore, Md. : 1950)
Burt BM,Plitas G,Zhao Z,Bamboat ZM,Nguyen HM,Dupont B,DeMatteo RP
11-9994 was used in Flow cytometry/Cell sorting to conclude that the human liver contains NK cell subsets which have reduced effector function, but under appropriate inflammatory conditions become potent killers.
Sat Aug 01 00:00:00 EDT 2009