Search Thermo Fisher Scientific
Immunogen sequence: MDAPHSKAA LDSINELPEN ILLELFTHVP ARQLLLNCRL VCSLWRDLID LMTLWKRKCL REGFITKDWD QPVADWKIFY FLRSLHRNLL RNPCAEEDMF AWQIDFNGGD RWKVESLPGA HGTDFPDPKV KKYFVTSYEM CLKSQLVDLV AEGYWEELLD TFRPDIVVKD WFAARADCGC TYQLKVQLAS ADYFVLASFE PPPVTIQQWN NATWTEVSYT FSDYPRGVRY ILFQHGGRDT QYWAGWYGPR VTNSSIVVSP KMTRNQASSE AQPGQKHGQE EAAQSPYRAV VQIF (1-293 aa encoded by BC020880)
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42 kDa) which may be involved in the control of the cell cycle.