Product References
Smc3 dosage regulates B cell transit through germinal centers and restricts their malignant transformation.
Nature immunology
Rivas MA,Meydan C,Chin CR,Challman MF,Kim D,Bhinder B,Kloetgen A,Viny AD,Teater MR,McNally DR,Doane AS,Béguelin W,Fernández MTC,Shen H,Wang X,Levine RL,Chen Z,Tsirigos A,Elemento O,Mason CE,Melnick AM
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Niche-specific MHC II and PD-L1 regulate CD4+CD8αα+ intraepithelial lymphocyte differentiation.
The Journal of experimental medicine
Moon S,Park Y,Hyeon S,Kim YM,Kim JH,Kim H,Park S,Lee KJ,Koo BK,Ha SJ,Lee SW
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Apr 05 00:00:00 EDT 2021
Aggravation of hepatic ischemia‑reperfusion injury with increased inflammatory cell infiltration is associated with the TGF‑β/Smad3 signaling pathway.
Molecular medicine reports
Li H,Shen X,Tong Y,Ji T,Feng Y,Tang Y,Mai R,Ye J,Que T,Luo X
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Immunohistochemistry
Sun Aug 01 00:00:00 EDT 2021
Brd4 modulates diet-induced obesity via PPARγ-dependent Gdf3 expression in adipose tissue macrophages.
JCI insight
Hu X,Dong X,Li G,Chen Y,Chen J,He X,Sun H,Kim DH,Kemper JK,Chen LF
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Thu Apr 08 00:00:00 EDT 2021
Differentiation of fetal hematopoietic stem cells requires ARID4B to restrict autocrine KITLG/KIT-Src signaling.
Cell reports
Young IC,Wu B,Andricovich J,Chuang ST,Li R,Tzatsos A,Wu RC,Wu MY
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Nov 23 00:00:00 EST 2021
Erythritol Ameliorates Small Intestinal Inflammation Induced by High-Fat Diets and Improves Glucose Tolerance.
International journal of molecular sciences
Kawano R,Okamura T,Hashimoto Y,Majima S,Senmaru T,Ushigome E,Asano M,Yamazaki M,Takakuwa H,Sasano R,Nakanishi N,Hamaguchi M,Fukui M
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon May 24 00:00:00 EDT 2021
Expansion of an Unusual Virtual Memory CD8+ Subpopulation Bearing Vα3.2 TCR in Themis-Deficient Mice.
Frontiers in immunology
Prasad M,Wojciech L,Brzostek J,Hu J,Chua YL,Tung DWH,Yap J,Rybakin V,Gascoigne NRJ
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Thu Sep 23 00:00:00 EDT 2021
The subunits of IL-12, originating from two distinct cells, can functionally synergize to protect against pathogen dissemination in vivo.
Cell reports
Gerber AN,Abdi K,Singh NJ
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Oct 12 00:00:00 EDT 2021
Ddb1 Is Essential for the Expansion of CD4+ Helper T Cells by Regulating Cell Cycle Progression and Cell Death.
Frontiers in immunology
Yang L,Chen W,Li L,Xiao Y,Fan S,Zhang Q,Xia T,Li M,Hong Y,Zhao T,Li Q,Liu WH,Xiao N
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Dec 20 00:00:00 EST 2021
Abraxane-induced bone marrow CD11b+ myeloid cell depletion in tumor-bearing mice is visualized by μPET-CT with 64Cu-labeled anti-CD11b and prevented by anti-CSF-1.
Theranostics
Cao Q,Huang Q,Wang YA,Li C
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Fri Jul 23 00:00:00 EDT 2021
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2020
Proenkephalin+ regulatory T cells expanded by ultraviolet B exposure maintain skin homeostasis with a healing function.
Proceedings of the National Academy of Sciences of the United States of America
Shime H,Odanaka M,Tsuiji M,Matoba T,Imai M,Yasumizu Y,Uraki R,Minohara K,Watanabe M,Bonito AJ,Fukuyama H,Ohkura N,Sakaguchi S,Morita A,Yamazaki S
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Aug 25 00:00:00 EDT 2020
Interleukin 35 Delays Hindlimb Ischemia-Induced Angiogenesis Through Regulating ROS-Extracellular Matrix but Spares Later Regenerative Angiogenesis.
Frontiers in immunology
Fu H,Sun Y,Shao Y,Saredy J,Cueto R,Liu L,Drummer C,Johnson C,Xu K,Lu Y,Li X,Meng S,Xue ER,Tan J,Jhala NC,Yu D,Zhou Y,Bayless KJ,Yu J,Rogers TJ,Hu W,Snyder NW,Sun J,Qin X,Jiang X,Wang H,Yang X
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Fri Jul 09 00:00:00 EDT 2021
The frequency of follicular T helper cells differs in acute and chronic neuroinflammation.
Scientific reports
Baniahmad A,Birkner K,Görg J,Loos J,Zipp F,Wasser B,Bittner S
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Nov 24 00:00:00 EST 2020
Long-term ex vivo haematopoietic-stem-cell expansion allows nonconditioned transplantation.
Nature
Wilkinson AC,Ishida R,Kikuchi M,Sudo K,Morita M,Crisostomo RV,Yamamoto R,Loh KM,Nakamura Y,Watanabe M,Nakauchi H,Yamazaki S
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
Focal adhesion kinase (FAK) deficiency in mononuclear phagocytes alters murine breast tumor progression.
American journal of cancer research
Llewellyn RA,Gutknecht MF,Thomas KS,Conaway MR,Bouton AH
13-0454 was used in Flow cytometry/Cell sorting to determine that focal adhesion kinase expression in this cell population influences primary breast tumour initiation and growth.
Wed Sep 30 00:00:00 EDT 2020
The H3K4 methyltransferase Setd1b is essential for hematopoietic stem and progenitor cell homeostasis in mice.
eLife
Schmidt K,Zhang Q,Tasdogan A,Petzold A,Dahl A,Arneth BM,Slany R,Fehling HJ,Kranz A,Stewart AF,Anastassiadis K
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Jun 19 00:00:00 EDT 2018
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus.
eLife
Moretti FA,Klapproth S,Ruppert R,Margraf A,Weber J,Pick R,Scheiermann C,Sperandio M,Fässler R,Moser M
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Thu Sep 06 00:00:00 EDT 2018
Elimination of intravascular thrombi prevents early mortality and reduces gliosis in hyper-inflammatory experimental cerebral malaria.
Journal of neuroinflammation
Wilson KD,Ochoa LF,Solomon OD,Pal R,Cardona SM,Carpio VH,Keiser PH,Cardona AE,Vargas G,Stephens R
13-0454 was used in Immunohistochemistry on frozen tissues to study the congestion of vasculature with leukocytes and fibrinogen,and the relationship of glial cell activation to congested vessels in the brains of Plasmodium chabaudi-infected, IL-10 KO mice.
Mon Jun 04 00:00:00 EDT 2018
Elimination of intravascular thrombi prevents early mortality and reduces gliosis in hyper-inflammatory experimental cerebral malaria.
Journal of neuroinflammation
Wilson KD,Ochoa LF,Solomon OD,Pal R,Cardona SM,Carpio VH,Keiser PH,Cardona AE,Vargas G,Stephens R
13-0454 was used in Immunohistochemistry on frozen tissues to study the congestion of vasculature with leukocytes and fibrinogen,and the relationship of glial cell activation to congested vessels in the brains of Plasmodium chabaudi-infected, IL-10 KO mice.
Mon Jun 04 00:00:00 EDT 2018
Spred1 Safeguards Hematopoietic Homeostasis against Diet-Induced Systemic Stress.
Cell stem cell
Tadokoro Y,Hoshii T,Yamazaki S,Eto K,Ema H,Kobayashi M,Ueno M,Ohta K,Arai Y,Hara E,Harada K,Oshima M,Oshima H,Arai F,Yoshimura A,Nakauchi H,Hirao A
13-0454-82 was used in Flow Cytometry to demonstrate the involvement of diet-induced stress on hematopoietic stem cell self-renewal.
Thu May 03 00:00:00 EDT 2018
Myeloid progenitor cluster formation drives emergency and leukaemic myelopoiesis.
Nature
Hérault A,Binnewies M,Leong S,Calero-Nieto FJ,Zhang SY,Kang YA,Wang X,Pietras EM,Chu SH,Barry-Holson K,Armstrong S,Göttgens B,Passegué E
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Immunofluorescence
Thu Apr 06 00:00:00 EDT 2017
Egr2 and 3 control adaptive immune responses by temporally uncoupling expansion from T cell differentiation.
The Journal of experimental medicine
Miao T,Symonds ALJ,Singh R,Symonds JD,Ogbe A,Omodho B,Zhu B,Li S,Wang P
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Jun 05 00:00:00 EDT 2017
Recipient bone marrow assimilates the myeloid/lymphoid reconstitution of distinct fetal hematopoietic stem cells.
Oncotarget
Guo XL,Chu L,Ke F,Mu LL,Li Z,Cai JJ,Li HF,Hong DL
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Dec 12 00:00:00 EST 2017
Pneumocystis-Driven Inducible Bronchus-Associated Lymphoid Tissue Formation Requires Th2 and Th17 Immunity.
Cell reports
Eddens T,Elsegeiny W,Garcia-Hernadez ML,Castillo P,Trevejo-Nunez G,Serody K,Campfield BT,Khader SA,Chen K,Rangel-Moreno J,Kolls JK
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Mar 28 00:00:00 EDT 2017
Intratumoral modulation of the inducible co-stimulator ICOS by recombinant oncolytic virus promotes systemic anti-tumour immunity.
Nature communications
Zamarin D,Holmgaard RB,Ricca J,Plitt T,Palese P,Sharma P,Merghoub T,Wolchok JD,Allison JP
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2017
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Thu Oct 06 00:00:00 EDT 2016
The Deubiquitinase OTULIN Is an Essential Negative Regulator of Inflammation and Autoimmunity.
Cell
Damgaard RB,Walker JA,Marco-Casanova P,Morgan NV,Titheradge HL,Elliott PR,McHale D,Maher ER,McKenzie ANJ,Komander D
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Thu Aug 25 00:00:00 EDT 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
LTβR controls thymic portal endothelial cells for haematopoietic progenitor cell homing and T-cell regeneration.
Nature communications
Shi Y,Wu W,Chai Q,Li Q,Hou Y,Xia H,Ren B,Xu H,Guo X,Jin C,Lv M,Wang Z,Fu YX,Zhu M
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Immunofluorescence
Fri Aug 05 00:00:00 EDT 2016
Atypical chemokine receptor 1 deficiency reduces atherogenesis in ApoE-knockout mice.
Cardiovascular research
Wan W,Liu Q,Lionakis MS,Marino AP,Anderson SA,Swamydas M,Murphy PM
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Axitinib increases the infiltration of immune cells and reduces the suppressive capacity of monocytic MDSCs in an intracranial mouse melanoma model.
Oncoimmunology
Du Four S,Maenhout SK,De Pierre K,Renmans D,Niclou SP,Thielemans K,Neyns B,Aerts JL
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Flow-dependent epigenetic DNA methylation regulates endothelial gene expression and atherosclerosis.
The Journal of clinical investigation
Dunn J,Qiu H,Kim S,Jjingo D,Hoffman R,Kim CW,Jang I,Son DJ,Kim D,Pan C,Fan Y,Jordan IK,Jo H
13-0454 was used in Immunohistochemistry on frozen tissues to determine that disturbed blood flow regulates genome-wide DNA methylation patterns in a DNA methyltransferase-dependent manner, altering endothelial gene expression and inducing atherosclerosis.
Tue Jul 01 00:00:00 EDT 2014
WASH is required for the differentiation commitment of hematopoietic stem cells in a c-Myc-dependent manner.
The Journal of experimental medicine
Xia P,Wang S,Huang G,Zhu P,Li M,Ye B,Du Y,Fan Z
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Mon Sep 22 00:00:00 EDT 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Wed Jun 10 00:00:00 EDT 2015
Mutual expression of the transcription factors Runx3 and ThPOK regulates intestinal CD4⁺ T cell immunity.
Nature immunology
Reis BS,Rogoz A,Costa-Pinto FA,Taniuchi I,Mucida D
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
In vivo generation of transplantable human hematopoietic cells from induced pluripotent stem cells.
Blood
Amabile G,Welner RS,Nombela-Arrieta C,D'Alise AM,Di Ruscio A,Ebralidze AK,Kraytsberg Y,Ye M,Kocher O,Neuberg DS,Khrapko K,Silberstein LE,Tenen DG
13-0454 was used in Flow cytometry/Cell sorting to develop an in vivo system in which human induced pluripotent stem cells differentiate within teratomas to derive functional myeloid and lymphoid cells.
Thu Feb 21 00:00:00 EST 2013
Distinct functions of chemokine receptor axes in the atherogenic mobilization and recruitment of classical monocytes.
EMBO molecular medicine
Soehnlein O,Drechsler M,Döring Y,Lievens D,Hartwig H,Kemmerich K,Ortega-Gómez A,Mandl M,Vijayan S,Projahn D,Garlichs CD,Koenen RR,Hristov M,Lutgens E,Zernecke A,Weber C
13-0454 was used in Immunohistochemistry on frozen tissues to investigate the differential role of classical and non-classical leukocytes in atheroprogression under high-fat diet, showing that CXCL1 and CCR1/CCR5 are important in the mobilisation and recruitment of classical monocytes in atheroprogression.
Fri Mar 01 00:00:00 EST 2013
Arrayed lentiviral barcoding for quantification analysis of hematopoietic dynamics.
Stem cells (Dayton, Ohio)
Grosselin J,Sii-Felice K,Payen E,Chretien S,Tronik-Le Roux D,Leboulch P
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
Roles of chemokine receptor CX3CR1 in maintaining murine bone homeostasis through the regulation of both osteoblasts and osteoclasts.
Journal of cell science
Hoshino A,Ueha S,Hanada S,Imai T,Ito M,Yamamoto K,Matsushima K,Yamaguchi A,Iimura T
13-0454 was used in Flow cytometry/Cell sorting to analyse the bone phenotypes of CX3CR1-deficient mice to further assess the roles of chemokine receptors in bone metabolism.
Fri Feb 15 00:00:00 EST 2013
The cardioprotective protein apolipoprotein A1 promotes potent anti-tumorigenic effects.
The Journal of biological chemistry
Zamanian-Daryoush M,Lindner D,Tallant TC,Wang Z,Buffa J,Klipfell E,Parker Y,Hatala D,Parsons-Wingerter P,Rayman P,Yusufishaq MSS,Fisher EA,Smith JD,Finke J,DiDonato JA,Hazen SL
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Flow Cytometry
Fri Jul 19 00:00:00 EDT 2013
The specialized unfolded protein response of B lymphocytes: ATF6α-independent development of antibody-secreting B cells.
Molecular immunology
Aragon IV,Barrington RA,Jackowski S,Mori K,Brewer JW
13-0454 was used in Flow cytometry/Cell sorting to demonstrate that a specialised B cell UPR relies exclusively on the IRE1-XBP1 pathway to remodel the ER and expand secretory capacity.
Sun Jul 01 00:00:00 EDT 2012
The earliest intrathymic precursors of CD8α(+) thymic dendritic cells correspond to myeloid-type double-negative 1c cells.
European journal of immunology
Luche H,Ardouin L,Teo P,See P,Henri S,Merad M,Ginhoux F,Malissen B
13-0454 was used in Flow cytometry/Cell sorting to provide evidence that myeloid-restricted progenitors generate the whole constellation of DCs present in the body, including the thymus.
Mon Aug 01 00:00:00 EDT 2011
Cellular competition independent of BAFF/B lymphocyte stimulator results in low frequency of an autoreactive clonotype in mature polyclonal B cell compartments.
Journal of immunology (Baltimore, Md. : 1950)
Nikbakht N,Migone TS,Ward CP,Manser T
13-0454 was used in Flow cytometry/Cell sorting to investigate the role of the peripheral B cell pro-survival cytokine BAFF/BLyS in the regulation of immunological tolerance and autoreactive cells.
Fri Jul 01 00:00:00 EDT 2011
Initial clonal expansion of germinal center B cells takes place at the perimeter of follicles.
Immunity
Coffey F,Alabyev B,Manser T
13-0454 was used in Immunohistochemistry on frozen tissues to study the proliferation, differentiation, and microenvironmental locale in populations of pre-germinal centre B cells responding to antigen.
Fri Apr 17 00:00:00 EDT 2009
Tracking epitope-specific T cells.
Nature protocols
Moon JJ,Chu HH,Hataye J,Pagán AJ,Pepper M,McLachlan JB,Zell T,Jenkins MK
13-0454 was used in Flow cytometry/Cell sorting to track epitope-specific T cells.
Tue Jun 30 00:00:00 EDT 2009
The lupus susceptibility locus Sle1 breaches peripheral B cell tolerance at the antibody-forming cell and germinal center checkpoints.
Journal of immunology (Baltimore, Md. : 1950)
Vuyyuru R,Mohan C,Manser T,Rahman ZS
13-0454 was used in Magnetic cell separation to show that Sle1 perturbs the action of peripheral tolerance checkpoints operative on antinuclear Ag B cells in both the AFC and GC pathways in a cell autonomous fashion.
Sun Nov 01 00:00:00 EDT 2009
In vivo equilibrium of proinflammatory IL-17+ and regulatory IL-10+ Foxp3+ RORgamma t+ T cells.
The Journal of experimental medicine
Lochner M,Peduto L,Cherrier M,Sawa S,Langa F,Varona R,Riethmacher D,Si-Tahar M,Di Santo JP,Eberl G
13-0454 was used in Flow cytometry/Cell sorting to study how potentially antagonistic pro-inflammatory IL-17-producing and regulatory Foxp3(+) RORgamma t(+) T cells coexist and are tightly controlled.
Mon Jun 09 00:00:00 EDT 2008
IL-7 specifies B cell fate at the common lymphoid progenitor to pre-proB transition stage by maintaining early B cell factor expression.
Journal of immunology (Baltimore, Md. : 1950)
Kikuchi K,Kasai H,Watanabe A,Lai AY,Kondo M
13-0454 was used in Flow cytometry/Cell sorting to investigate when IL-7 stimulation is necessary in hemato-/lymphopoiesis in adult mice, showing that IL-7 specifies B cell fate at the common lymphoid progenitor to pre-proB transition stage by maintaining early B cell factor expression.
Tue Jul 01 00:00:00 EDT 2008
TGFbeta in the context of an inflammatory cytokine milieu supports de novo differentiation of IL-17-producing T cells.
Immunity
Veldhoen M,Hocking RJ,Atkins CJ,Locksley RM,Stockinger B
13-0454 was used in Flow cytometry/Cell sorting to investigate the generation of IL-17-producing T cells from naive CD4 T cells, showing that TGFbeta supports de novo differentiation of IL-17-producing T cells.
Wed Feb 01 00:00:00 EST 2006
Enhanced engraftment of hematopoietic stem/progenitor cells by the transient inhibition of an adaptor protein, Lnk.
Blood
Takizawa H,Kubo-Akashi C,Nobuhisa I,Kwon SM,Iseki M,Taga T,Takatsu K,Takaki S
13-0454 was used in Flow cytometry/Cell sorting to investigate the potential of Lnk for augmented engraftment of HSCs.
Sat Apr 01 00:00:00 EST 2006
Developmental switch of mouse hematopoietic stem cells from fetal to adult type occurs in bone marrow after birth.
Proceedings of the National Academy of Sciences of the United States of America
Kikuchi K,Kondo M
13-0454 was used in Flow cytometry/Cell sorting to demonstrate how foetal haematopoietic stem cells may acquire adult characteristics between 1 and 2 weeks after birth in mouse bone marrow.
Tue Nov 21 00:00:00 EST 2006
Plk1 regulates activation of the anaphase promoting complex by phosphorylating and triggering SCFbetaTrCP-dependent destruction of the APC Inhibitor Emi1.
Molecular biology of the cell
Hansen DV,Loktev AV,Ban KH,Jackson PK
Published figure using CD45.2 monoclonal antibody (Product # 13-0454-82) in Western Blot
Wed Dec 01 00:00:00 EST 2004