Product References
Mechanical Compression by Simulating Orthodontic Tooth Movement in an In Vitro Model Modulates Phosphorylation of AKT and MAPKs via TLR4 in Human Periodontal Ligament Cells.
International journal of molecular sciences
Roth CE,Craveiro RB,Niederau C,Malyaran H,Neuss S,Jankowski J,Wolf M
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Fri Jul 22 00:00:00 EDT 2022
Lys694Arg polymorphism leads to blunted responses to LPS by interfering TLR4 with recruitment of MyD88.
Innate immunity
Yang Y,Hu Y,Zhou Y,Liang T,Tang H,Ju H,Shi Q,Fang H
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Western Blot
Sun Aug 01 00:00:00 EDT 2021
Inflammatory signaling in dengue-infected platelets requires translation and secretion of nonstructural protein 1.
Blood advances
Quirino-Teixeira AC,Rozini SV,Barbosa-Lima G,Coelho DR,Carneiro PH,Mohana-Borges R,Bozza PT,Hottz ED
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Tue May 12 00:00:00 EDT 2020
Grindelia squarrosa Extract and Grindelic Acid Modulate Pro-inflammatory Functions of Respiratory Epithelium and Human Macrophages.
Frontiers in pharmacology
Gierlikowska B,Filipek A,Gierlikowski W,Kania D,Stefańska J,Demkow U,Kiss AK
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Sat Feb 06 00:00:00 EST 2021
Bacterial lipopolysaccharide and antimicrobial LL-37 enhance ICAM-1 expression and NF-κB p65 phosphorylation in senescent endothelial cells.
International journal of molecular medicine
Suzuki K,Ohkuma M,Nagaoka I
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
Toll‑Like receptor 4 promotes the phosphorylation of CRMP2 via the activation of Rho‑kinase in MCAO rats.
Molecular medicine reports
Li XB,Ding MX,Ding CL,Li LL,Feng J,Yu XJ
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Sun Jul 01 00:00:00 EDT 2018
n-butanol extract from Folium isatidis inhibits the lipopolysaccharide-induced downregulation of CXCR1 and CXCR2 on human neutrophils.
Molecular medicine reports
Wu B,Wang L,Jiang L,Dong L,Xu F,Lu Y,Jin J,Wang Z,Liang G,Shan X
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Butyrate upregulates the TLR4 expression and the phosphorylation of MAPKs and NK-κB in colon cancer cell in vitro.
Oncology letters
Xiao T,Wu S,Yan C,Zhao C,Jin H,Yan N,Xu J,Wu Y,Li C,Shao Q,Xia S
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
Bushfire smoke is pro-inflammatory and suppresses macrophage phagocytic function.
Scientific reports
Hamon R,Tran HB,Roscioli E,Ween M,Jersmann H,Hodge S
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Fri Sep 07 00:00:00 EDT 2018
Altered Toll-like receptor expression and function in HPV-associated oropharyngeal carcinoma.
Oncotarget
Tobouti PL,Bolt R,Radhakrishnan R,de Sousa SCOM,Hunter KD
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Tue Jan 02 00:00:00 EST 2018
A low dose lipid infusion is sufficient to induce insulin resistance and a pro-inflammatory response in human subjects.
PloS one
Liang H,Lum H,Alvarez A,Garduno-Garcia JJ,Daniel BJ,Musi N
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Fri Jul 27 00:00:00 EDT 2018
Platelet proteome reveals novel pathways of platelet activation and platelet-mediated immunoregulation in dengue.
PLoS pathogens
Trugilho MRO,Hottz ED,Brunoro GVF,Teixeira-Ferreira A,Carvalho PC,Salazar GA,Zimmerman GA,Bozza FA,Bozza PT,Perales J
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Mon May 01 00:00:00 EDT 2017
Pulmonary immune responses to Mycobacterium tuberculosis in exposed individuals.
PloS one
Herzmann C,Ernst M,Lange C,Stenger S,Kaufmann SHE,Reiling N,Schaberg T,van der Merwe L,Maertzdorf J
13-9917 was used in Flow cytometry/Cell sorting to investigate the correlation between interferon-(IFN)-γ release assays and pulmonary immune responses in tuberculosis contacts in Germany.
Mon Dec 04 00:00:00 EST 2017
Picfeltarraenin IA inhibits lipopolysaccharide-induced inflammatory cytokine production by the nuclear factor-κB pathway in human pulmonary epithelial A549 cells.
Oncology letters
Shi R,Wang Q,Ouyang Y,Wang Q,Xiong X
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Mon Feb 01 00:00:00 EST 2016
Lipopolysaccharide induces SBD-1 expression via the P38 MAPK signaling pathway in ovine oviduct epithelial cells.
Lipids in health and disease
Li Q,Bao F,Zhi D,Liu M,Yan Q,Zheng X,Ren L,Cong S,Li Y,Cao G
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Thu Aug 11 00:00:00 EDT 2016
Lipopolysaccharide induces SBD-1 expression via the P38 MAPK signaling pathway in ovine oviduct epithelial cells.
Lipids in health and disease
Li Q,Bao F,Zhi D,Liu M,Yan Q,Zheng X,Ren L,Cong S,Li Y,Cao G
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Thu Aug 11 00:00:00 EDT 2016
TLR4 Expression Is Associated with Left Ventricular Dysfunction in Patients Undergoing Coronary Artery Bypass Surgery.
PloS one
Avlas O,Bragg A,Fuks A,Nicholson JD,Farkash A,Porat E,Aravot D,Levy-Drummer RS,Cohen C,Shainberg A,Arad M,Hochhauser E
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2016
TLR4 Expression Is Associated with Left Ventricular Dysfunction in Patients Undergoing Coronary Artery Bypass Surgery.
PloS one
Avlas O,Bragg A,Fuks A,Nicholson JD,Farkash A,Porat E,Aravot D,Levy-Drummer RS,Cohen C,Shainberg A,Arad M,Hochhauser E
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2016
The COP II adaptor protein TMED7 is required to initiate and mediate the delivery of TLR4 to the plasma membrane.
Science signaling
Liaunardy-Jopeace A,Bryant CE,Gay NJ
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Immunofluorescence
Tue Jul 29 00:00:00 EDT 2014
The COP II adaptor protein TMED7 is required to initiate and mediate the delivery of TLR4 to the plasma membrane.
Science signaling
Liaunardy-Jopeace A,Bryant CE,Gay NJ
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Immunofluorescence
Tue Jul 29 00:00:00 EDT 2014
Biologically active polymers from spontaneous carotenoid oxidation: a new frontier in carotenoid activity.
PloS one
Johnston JB,Nickerson JG,Daroszewski J,Mogg TJ,Burton GW
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Immunocytochemistry
Thu Jun 25 00:00:00 EDT 2015
Body fluid exosomes promote secretion of inflammatory cytokines in monocytic cells via Toll-like receptor signaling.
The Journal of biological chemistry
Bretz NP,Ridinger J,Rupp AK,Rimbach K,Keller S,Rupp C,Marmé F,Umansky L,Umansky V,Eigenbrod T,Sammar M,Altevogt P
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Fri Dec 20 00:00:00 EST 2013
Body fluid exosomes promote secretion of inflammatory cytokines in monocytic cells via Toll-like receptor signaling.
The Journal of biological chemistry
Bretz NP,Ridinger J,Rupp AK,Rimbach K,Keller S,Rupp C,Marmé F,Umansky L,Umansky V,Eigenbrod T,Sammar M,Altevogt P
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Fri Dec 20 00:00:00 EST 2013
Increased frequencies of pulmonary regulatory T-cells in latent Mycobacterium tuberculosis infection.
The European respiratory journal
Herzmann C,Ernst M,Ehlers S,Stenger S,Maertzdorf J,Sotgiu G,Lange C
13-9917 was used in Flow cytometry/Cell sorting to study how latent infection with M. tuberculosis, as defined by a positive M. tuberculosis-specific IGRA response on cells from the peripheral blood, is characterised by an increased frequency of Treg cells in the BAL.
Sat Dec 01 00:00:00 EST 2012
TLR4 protein contributes to cigarette smoke-induced matrix metalloproteinase-1 (MMP-1) expression in chronic obstructive pulmonary disease.
The Journal of biological chemistry
Geraghty P,Dabo AJ,D'Armiento J
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in ELISA
Fri Aug 26 00:00:00 EDT 2011
TLR4 protein contributes to cigarette smoke-induced matrix metalloproteinase-1 (MMP-1) expression in chronic obstructive pulmonary disease.
The Journal of biological chemistry
Geraghty P,Dabo AJ,D'Armiento J
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in ELISA
Fri Aug 26 00:00:00 EDT 2011
Candesartan inhibits Toll-like receptor expression and activity both in vitro and in vivo.
Atherosclerosis
Dasu MR,Riosvelasco AC,Jialal I
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2009
Postnatal development of monocyte cytokine responses to bacterial lipopolysaccharide.
Pediatric research
Yerkovich ST,Wikström ME,Suriyaarachchi D,Prescott SL,Upham JW,Holt PG
13-9917 was used in Flow cytometry/Cell sorting to investigate the capacity of mononuclear cells to secrete inflammatory cytokines in response to interferon-gamma priming and lipopolysaccharide stimulation.
Thu Nov 01 00:00:00 EDT 2007
Human heat shock protein 60 stimulates vascular smooth muscle cell proliferation through Toll-like receptors 2 and 4.
Microbes and infection
de Graaf R,Kloppenburg G,Kitslaar PJ,Bruggeman CA,Stassen F
Thu Jun 01 00:00:00 EDT 2006
Respiratory epithelial cells require Toll-like receptor 4 for induction of human beta-defensin 2 by lipopolysaccharide.
Respiratory research
MacRedmond R,Greene C,Taggart CC,McElvaney N,O'Neill S
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Wed Oct 12 00:00:00 EDT 2005
TLR9 is localized in the endoplasmic reticulum prior to stimulation.
Journal of immunology (Baltimore, Md. : 1950)
Leifer CA,Kennedy MN,Mazzoni A,Lee C,Kruhlak MJ,Segal DM
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Immunofluorescence
Thu Jul 15 00:00:00 EDT 2004
Differential regulation of inflammatory cytokine secretion by human dendritic cells upon Chlamydia trachomatis infection.
Infection and immunity
Gervassi A,Alderson MR,Suchland R,Maisonneuve JF,Grabstein KH,Probst P
Published figure using CD284 (TLR4) monoclonal antibody (Product # 13-9917-82) in Neutralization
Wed Dec 01 00:00:00 EST 2004
Expression and involvement of Toll-like receptors (TLR)2, TLR4, and CD14 in monocyte TNF-alpha production induced by lipopolysaccharides from Neisseria meningitidis.
Medical science monitor : international medical journal of experimental and clinical research
Mirlashari MR,Lyberg T
Fri Aug 01 00:00:00 EDT 2003