Search Thermo Fisher Scientific
Immunogen sequence: FGEPHIPSR LPPLPLHIRI QQALTSPLPM TPILEGSHRA HSLLFENSDS FSEDSSTLGR TRSLPITIEM LKVPDDEEEE EQTCPSTFSE EMTPTSVIPK LPQCLREEEE KESDSDSEGP IQYRDEEDED ESYQSALANK VKRKDTLAMK LNHRPSEPEL NLNSWPCKSK EEWNEIRHQI GNTLIRRLSQ RPTPEELEQR NILQPKNEAD RQAEKREIKR RLTRKLSQRP TVAELLARKI LRFNEYVEVT DAQDYDRRAD KPWTKLTPAD KAAIRKELNE FKSSEMEVHE ESKHFTRYHR P (403-702 aa encoded by BC029266)
This gene encodes a member of the phosphatase and actin regulator (PHACTR) family. Other PHACTR family members have been shown to inhibit protein phosphatase 1 (PP1) activity, and the homolog of this gene in the mouse has been shown to interact with actin and PP1. Multiple transcript variants encoding different isoforms have been found for this gene.
Protein Aliases: Humdy; Phosphatase and actin regulator 4; Protein Humpty dumpty; protein phosphatase 1, regulatory subunit 124
Gene Aliases: 3110001B12Rik; AI527228; AW495572; C330013F19Rik; Kiaa4120; mKIAA4120; N28169; PHACTR4; PPP1R124; PRO2963
UniProt ID: (Human) Q8IZ21, (Mouse) Q501J7
Entrez Gene ID: (Human) 65979, (Mouse) 100169, (Rat) 682920