Product References
BRAF D594A mutation defines a unique biological and immuno-modulatory subgroup associated with functional CD8+ T cell infiltration in colorectal cancer.
Journal of translational medicine
Li W,Zhao C,Li W,Gong Y,Ma K,Lu Y,Liu X,Zhang L,Guo F
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Wed Oct 18 00:00:00 EDT 2023
Sequence-based drug design as a concept in computational drug design.
Nature communications
Chen L,Fan Z,Chang J,Yang R,Hou H,Guo H,Zhang Y,Yang T,Zhou C,Sui Q,Chen Z,Zheng C,Hao X,Zhang K,Cui R,Zhang Z,Ma H,Ding Y,Zhang N,Lu X,Luo X,Jiang H,Zhang S,Zheng M
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Fri Jul 14 00:00:00 EDT 2023
Extracellular acidosis restricts one-carbon metabolism and preserves T cell stemness.
Nature metabolism
Cheng H,Qiu Y,Xu Y,Chen L,Ma K,Tao M,Frankiw L,Yin H,Xie E,Pan X,Du J,Wang Z,Zhu W,Chen L,Zhang L,Li G
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2023
TIM3 Expression in Anaplastic-Thyroid-Cancer-Infiltrating Macrophages: An Emerging Immunotherapeutic Target.
Biology
Palacios LM,Peyret V,Viano ME,Geysels RC,Chocobar YA,Volpini X,Pellizas CG,Nicola JP,Motran CC,Rodriguez-Galan MC,Fozzatti L
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Thu Nov 03 00:00:00 EDT 2022
Breast cancer cell-derived extracellular vesicles promote CD8+ T cell exhaustion via TGF-β type II receptor signaling.
Nature communications
Xie F,Zhou X,Su P,Li H,Tu Y,Du J,Pan C,Wei X,Zheng M,Jin K,Miao L,Wang C,Meng X,van Dam H,Ten Dijke P,Zhang L,Zhou F
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
Functional assessment of the cell-autonomous role of NADase CD38 in regulating CD8+ T cell exhaustion.
iScience
Ma K,Sun L,Shen M,Zhang X,Xiao Z,Wang J,Liu X,Jiang K,Xiao-Feng Qin F,Guo F,Zhang B,Zhang L
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Fri May 20 00:00:00 EDT 2022
Mobilizing phospholipids on tumor plasma membrane implicates phosphatidylserine externalization blockade for cancer immunotherapy.
Cell reports
Wang W,Wu S,Cen Z,Zhang Y,Chen Y,Huang Y,Cillo AR,Prokopec JS,Quarato G,Vignali DAA,Stewart-Ornstein J,Li S,Lu B,Gong YN
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
Type 1 Cytotoxic T Cells Increase in Placenta after Intrauterine Inflammation.
Frontiers in immunology
Liu J,Liu Y,Panda S,Liu A,Lei J,Burd I
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Fri Dec 17 00:00:00 EST 2021
PTENα functions as an immune suppressor and promotes immune resistance in PTEN-mutant cancer.
Nature communications
Sun Y,Lu D,Yin Y,Song J,Liu Y,Hao W,Qi F,Zhang G,Zhang X,Liu L,Lin Z,Liang H,Zhao X,Jin Y,Yin Y
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Thu Aug 26 00:00:00 EDT 2021
A Fas-4-1BB fusion protein converts a death to a pro-survival signal and enhances T cell therapy.
The Journal of experimental medicine
Oda SK,Anderson KG,Ravikumar P,Bonson P,Garcia NM,Jenkins CM,Zhuang S,Daman AW,Chiu EY,Bates BM,Greenberg PD
14-5870-82 was used in Flow Cytometry to develop enhanced adoptive T cell therapy using immunomodulatory fusion proteins for use in solid tumours.
Mon Dec 07 00:00:00 EST 2020
Inhibition of the Wnt/β-catenin pathway enhances antitumor immunity in ovarian cancer.
Therapeutic advances in medical oncology
Doo DW,Meza-Perez S,Londoño AI,Goldsberry WN,Katre AA,Boone JD,Moore DJ,Hudson CT,Betella I,McCaw TR,Gangrade A,Bao R,Luke JJ,Yang ES,Birrer MJ,Starenki D,Cooper SJ,Buchsbaum DJ,Norian LA,Randall TD,Arend RC
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Thu Apr 14 00:00:00 EDT 2022
Mechanisms of action of ruxolitinib in murine models of hemophagocytic lymphohistiocytosis.
Blood
Albeituni S,Verbist KC,Tedrick PE,Tillman H,Picarsic J,Bassett R,Nichols KE
14-5870-82 was used in Flow Cytometry to show that ruxolitinib operates through IFN-γ-dependent and -independent mechanisms to dampen HLH by targeting the deleterious effects of T cells and neutrophils, with the latter representing an unappreciated and understudied cell type that contributes to HLH pathogenesis.
Thu Jul 11 00:00:00 EDT 2019
Mapping the Lineage Relationship between CXCR5+ and CXCR5- CD4+ T Cells in HIV-Infected Human Lymph Nodes.
Cell reports
Del Alcazar D,Wang Y,He C,Wendel BS,Del Río-Estrada PM,Lin J,Ablanedo-Terrazas Y,Malone MJ,Hernandez SM,Frank I,Naji A,Reyes-Terán G,Jiang N,Su LF
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Tue Sep 17 00:00:00 EDT 2019
Down-Regulation of Tim-3 in Monocytes and Macrophages in Plasmodium Infection and Its Association with Parasite Clearance.
Frontiers in microbiology
Hou N,Jiang N,Zou Y,Piao X,Liu S,Li S,Chen Q
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Down-Regulation of Tim-3 in Monocytes and Macrophages in Plasmodium Infection and Its Association with Parasite Clearance.
Frontiers in microbiology
Hou N,Jiang N,Zou Y,Piao X,Liu S,Li S,Chen Q
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Down-Regulation of Tim-3 in Monocytes and Macrophages in Plasmodium Infection and Its Association with Parasite Clearance.
Frontiers in microbiology
Hou N,Jiang N,Zou Y,Piao X,Liu S,Li S,Chen Q
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
T-Cell Immunoglobulin- and Mucin-Domain-Containing Molecule 3 Signaling Blockade Improves Cell-Mediated Immunity Against Malaria.
The Journal of infectious diseases
Hou N,Zou Y,Piao X,Liu S,Wang L,Li S,Chen Q
Tue Nov 15 00:00:00 EST 2016
Growth and metastasis of lung adenocarcinoma is potentiated by BMP4-mediated immunosuppression.
Oncoimmunology
Chen L,Yi X,Goswami S,Ahn YH,Roybal JD,Yang Y,Diao L,Peng D,Peng D,Fradette JJ,Wang J,Byers LA,Kurie JM,Ullrich SE,Qin FX,Gibbons DL
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Tim-3 induces Th2-biased immunity and alternative macrophage activation during Schistosoma japonicum infection.
Infection and immunity
Hou N,Piao X,Liu S,Wu C,Chen Q
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
Tim-3 induces Th2-biased immunity and alternative macrophage activation during Schistosoma japonicum infection.
Infection and immunity
Hou N,Piao X,Liu S,Wu C,Chen Q
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
Combined immune checkpoint protein blockade and low dose whole body irradiation as immunotherapy for myeloma.
Journal for immunotherapy of cancer
Jing W,Gershan JA,Weber J,Tlomak D,McOlash L,Sabatos-Peyton C,Johnson BD
14587082 was used in flow cytometry to test if blocking several checkpoint receptors boosts anti-tumor immunity in a low-dose, lymphodepleting whole body radiation model
Fri Jan 23 00:00:00 EST 2015
Metastasis is regulated via microRNA-200/ZEB1 axis control of tumour cell PD-L1 expression and intratumoral immunosuppression.
Nature communications
Chen L,Gibbons DL,Goswami S,Cortez MA,Ahn YH,Byers LA,Zhang X,Yi X,Dwyer D,Lin W,Diao L,Wang J,Roybal J,Patel M,Ungewiss C,Peng D,Antonia S,Mediavilla-Varela M,Robertson G,Suraokar M,Welsh JW,Erez B,Wistuba II,Chen L,Peng D,Wang S,Ullrich SE,Heymach JV,Kurie JM,Qin FX
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Tue Oct 28 00:00:00 EDT 2014
T cell Ig mucin-3 promotes homeostasis of sepsis by negatively regulating the TLR response.
Journal of immunology (Baltimore, Md. : 1950)
Yang X,Jiang X,Chen G,Xiao Y,Geng S,Kang C,Zhou T,Li Y,Guo X,Xiao H,Hou C,Wang R,Lin Z,Li X,Feng J,Ma Y,Shen B,Li Y,Han G
14-5870 was used in Western Blotting to demonstrate a regulatory role for Tim-3 as part of a novel protective mechanism in sepsis victims.
Fri Mar 01 00:00:00 EST 2013
The influence of T cell Ig mucin-3 signaling on central nervous system autoimmune disease is determined by the effector function of the pathogenic T cells.
Journal of immunology (Baltimore, Md. : 1950)
Lee SY,Goverman JM
14-5870 was used in Flow cytometry/Cell sorting to analyse the role of Tim-3 signalling within various innate immune cells on the development of experimental autoimmune encephalomyelitis.
Wed May 15 00:00:00 EDT 2013
Transcutaneous vaccination via laser microporation.
Journal of controlled release : official journal of the Controlled Release Society
Weiss R,Hessenberger M,Kitzmüller S,Bach D,Weinberger EE,Krautgartner WD,Hauser-Kronberger C,Malissen B,Boehler C,Kalia YN,Thalhamer J,Scheiblhofer S
14-5870 was used in Flow cytometry/Cell sorting to investigate the potential of transcutaneous immunisation via laser-generated micropores for induction of specific immune responses.
Mon Sep 10 00:00:00 EDT 2012
CD8 T cells recruited early in mouse polyomavirus infection undergo exhaustion.
Journal of immunology (Baltimore, Md. : 1950)
Wilson JJ,Pack CD,Lin E,Frost EL,Albrecht JA,Hadley A,Hofstetter AR,Tevethia SS,Schell TD,Lukacher AE
14-5870 was used in Flow cytometry/Cell sorting to characterise virus-specific CD8 T cells recruited during persistent infection.
Tue May 01 00:00:00 EDT 2012
Exposure to nicotine adversely affects the dendritic cell system and compromises host response to vaccination.
Journal of immunology (Baltimore, Md. : 1950)
Nouri-Shirazi M,Guinet E
14-5870 was used in Flow cytometry/Cell sorting to investigate the causal relationship between nicotine-induced dendritic cell alterations and host response to vaccines.
Thu Mar 01 00:00:00 EST 2012
TIM-3 expression characterizes regulatory T cells in tumor tissues and is associated with lung cancer progression.
PloS one
Gao X,Zhu Y,Li G,Huang H,Zhang G,Wang F,Sun J,Yang Q,Zhang X,Lu B
14-5870 was used in Flow cytometry/Cell sorting to reveal a new role for TIM-3 as an important immune regulator in the tumour microenvironment.
Fri Jun 29 00:00:00 EDT 2012
T lymphocyte surface expression of exhaustion markers as biomarkers of the efficacy of chemotherapy for tuberculosis.
Tuberculosis (Edinburgh, Scotland)
Henao-Tamayo M,Irwin SM,Shang S,Ordway D,Orme IM
14-5870 was used in Flow cytometry/Cell sorting to identify that surface expression of PD-1, TIM-3 and KLRG-1 on T-lymphocytes changed with reduction of bacterial load in the lungs.
Fri Jul 01 00:00:00 EDT 2011
Tim-3 mediates phagocytosis of apoptotic cells and cross-presentation.
Blood
Nakayama M,Akiba H,Takeda K,Kojima Y,Hashiguchi M,Azuma M,Yagita H,Okumura K
Thu Apr 16 00:00:00 EDT 2009
Tim-3 mediates phagocytosis of apoptotic cells and cross-presentation.
Blood
Nakayama M,Akiba H,Takeda K,Kojima Y,Hashiguchi M,Azuma M,Yagita H,Okumura K
Thu Apr 16 00:00:00 EDT 2009
Phenotypic switching in Cryptococcus neoformans contributes to virulence by changing the immunological host response.
Infection and immunity
Guerrero A,Fries BC
Published figure using CD366 (TIM3) monoclonal antibody (Product # 14-5870-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2008