Product References
An ortholog of the Vasa intronic gene is required for small RNA-mediated translation repression in Chlamydomonas reinhardtii.
Proceedings of the National Academy of Sciences of the United States of America
Ma X,Ibrahim F,Kim EJ,Shaver S,Becker J,Razvi F,Cerny RL,Cerutti H
14-6995 was used in Western Blotting to show that VIG1 is an ancillary ribosomal component and plays a role in sRNA-mediated translation repression of polyribosomal transcripts.
Tue Jan 07 00:00:00 EST 2020
Baculovirus Surface Display of Zika Virus Envelope Protein Protects against Virus Challenge in Mouse Model.
Virologica Sinica
Luo D,Miao Y,Ke X,Tan Z,Hu C,Li P,Wang T,Zhang Y,Sun J,Liu Y,Wang H,Zheng Z
14-6995 was used in Western Blotting to demonstrate that the recombinant E protein displayed by baculovirus can be conveniently prepared and displayed good immunogenicity in immunized mice.
Thu Oct 01 00:00:00 EDT 2020
Autographa californica multiple nucleopolyhedrovirus ORF11 is essential for budded-virus production and occlusion-derived-virus envelopment.
Journal of virology
Tao XY,Choi JY,Kim WJ,An SB,Liu Q,Kim SE,Lee SH,Kim JH,Woo SD,Jin BR,Je YH
14-6995 was used in Western Blotting to study the ability of budded viruses to form in Ac11KO-transfected Sf9 cells.
Thu Jan 01 00:00:00 EST 2015
The production of KIR-Fc fusion proteins and their use in a multiplex HLA class I binding assay.
Journal of immunological methods
Hilton HG,Moesta AK,Guethlein LA,Blokhuis J,Parham P,Norman PJ
Thu Oct 01 00:00:00 EDT 2015
Critical assessment of influenza VLP production in Sf9 and HEK293 expression systems.
BMC biotechnology
Thompson CM,Petiot E,Mullick A,Aucoin MG,Henry O,Kamen AA
14-6995 was used in Western Blotting to examine two virus-like particle production systems for increasing influenza vaccine production rates.
Sat May 16 00:00:00 EDT 2015
Enhanced expression of full-length human cytomegalovirus fusion protein in non-swelling baculovirus-infected cells with a minimal fed-batch strategy.
PloS one
Patrone M,Carinhas N,Sousa MQ,Peixoto C,Ciferri C,Carfì A,Alves PM
14-6995 was used in Western Blotting to couple the exhaustive evaluation of the Baculovirus/insect cell expression system to an orthogonal screening for expression additives to produce full-length gB.
Tue Nov 04 00:00:00 EST 2014
The Autographa californica multiple nucleopolyhedrovirus ORF78 is essential for budded virus production and general occlusion body formation.
Journal of virology
Tao XY,Choi JY,Kim WJ,Lee JH,Liu Q,Kim SE,An SB,Lee SH,Woo SD,Jin BR,Je YH
14-6995 was used in Western Blotting to investigate the role of ac78 in the baculovirus life cycle.
Thu Aug 01 00:00:00 EDT 2013
Abrogation of contaminating RNA activity in HIV-1 Gag VLPs.
Virology journal
Valley-Omar Z,Meyers AE,Shephard EG,Williamson AL,Rybicki EP
14-6995 was used in Western Blotting to investigate methods to remove contaminating RNA from HIV-1 Gag virus like particle vaccine candidates.
Thu Oct 06 00:00:00 EDT 2011
Use of the piggyBac transposon to create HIV-1 gag transgenic insect cell lines for continuous VLP production.
BMC biotechnology
Lynch AG,Tanzer F,Fraser MJ,Shephard EG,Williamson AL,Rybicki EP
14-6995 was used in Western Blotting to showcase a novel method for continuous virus-like particle production as an alternative to the baculovirus system.
Wed Mar 31 00:00:00 EDT 2010
Purification of recombinant baculoviruses for gene therapy using membrane processes.
Gene therapy
Vicente T,Peixoto C,Carrondo MJ,Alves PM
14-6995 was used in Western Blotting to study the recovery and purification of rBVs aiming at injectable-grade virus batches for gene therapy trials.
Mon Jun 01 00:00:00 EDT 2009
Reduction of liver macrophage transduction by pseudotyping lentiviral vectors with a fusion envelope from Autographa californica GP64 and Sendai virus F2 domain.
BMC biotechnology
Markusic DM,van Til NP,Hiralall JK,Elferink RP,Seppen J
Wed Oct 07 00:00:00 EDT 2009
Lentivirus vector can be readministered to nasal epithelia without blocking immune responses.
Journal of virology
Sinn PL,Arias AC,Brogden KA,McCray PB
Published figure using Baculovirus Envelope gp64 monoclonal antibody (Product # 14-6995-82) in ELISA
Sat Nov 01 00:00:00 EDT 2008
Using double-stranded RNA to prevent in vitro and in vivo viral infections by recombinant baculovirus.
The Journal of biological chemistry
Valdes VJ,Sampieri A,Sepulveda J,Vaca L
14-6995-82 was used in Confocal Microscopy to show that dsRNAs from segments of two genes essential for baculovirus propagation prevent virus infection in vitro and in vivo.
Fri May 23 00:00:00 EDT 2003