Product References
Physioxic Culture of Chondrogenic Cells.
Methods in molecular biology (Clifton, N.J.)
Pattappa G,Markway BD,Docheva D,Johnstone B
14-9771-82 was used in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) to outline the methods for the expansion and differentiation of chondrogenic cells under physioxia using oxygen-controlled incubators and glove box environments, with the typical assays used for qualitative and quantitative assessment of chondrogenesis.
Mon Nov 14 00:00:00 EST 2022
Collagen X Is Dispensable for Hypertrophic Differentiation and Endochondral Ossification of Human iPSC-Derived Chondrocytes.
JBMR plus
Kamakura T,Jin Y,Nishio M,Nagata S,Fukuda M,Sun L,Kawai S,Toguchida J
Collagen X Is Dispensable for Hypertrophic Differentiation and Endochondral Ossification of Human iPSC-Derived Chondrocytes
Mon May 01 00:00:00 EDT 2023
Collagen X Is Dispensable for Hypertrophic Differentiation and Endochondral Ossification of Human iPSC-Derived Chondrocytes.
JBMR plus
Kamakura T,Jin Y,Nishio M,Nagata S,Fukuda M,Sun L,Kawai S,Toguchida J
Collagen X Is Dispensable for Hypertrophic Differentiation and Endochondral Ossification of Human iPSC-Derived Chondrocytes
Mon May 01 00:00:00 EDT 2023
Distinct Roles of Glutamine Metabolism in Benign and Malignant Cartilage Tumors With IDH Mutations.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Zhang H,Puviindran V,Nadesan P,Ding X,Shen L,Tang YJ,Tsushima H,Yahara Y,Ban GI,Zhang GF,Karner CM,Alman BA
14-9771-82 was used in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) to demonstrate that glutaminase-mediated glutamine metabolism played distinct roles in enchondromas and chondrosarcomas with IDH1 or IDH2 mutations.
Sun May 01 00:00:00 EDT 2022
The Effects of Fluvastatin on Indian Hedgehog Pathway in Endochondral Ossification.
Cartilage
Ishikawa M,Ishii T,Morikawa T,Iijima Y,Sueishi K
14-9771-82 was used in Immunohistochemistry to demonstrate that statins had an influence on Ihh, and that the hyperfunction of Ihh may prevent premature cell death caused by FGFR3-related chondrodysplasia through an indirect increase in the expression of PTHrP.
Wed Dec 01 00:00:00 EST 2021
Spatiotemporal distribution of thrombospondin-4 and -5 in cartilage during endochondral bone formation and repair.
Bone
Andrés Sastre E,Maly K,Zhu M,Witte-Bouma J,Trompet D,Böhm AM,Brachvogel B,van Nieuwenhoven CA,Maes C,van Osch GJVM,Zaucke F,Farrell E
14-9771-82 was used in Immunohistochemistry (Paraffin) to compare the spatiotemporal expression of thrombospondin-5 and thrombospondin-4 during postnatal bone formation and fracture healing.
Wed Sep 01 00:00:00 EDT 2021
Spatiotemporal distribution of thrombospondin-4 and -5 in cartilage during endochondral bone formation and repair.
Bone
Andrés Sastre E,Maly K,Zhu M,Witte-Bouma J,Trompet D,Böhm AM,Brachvogel B,van Nieuwenhoven CA,Maes C,van Osch GJVM,Zaucke F,Farrell E
14-9771-82 was used in Immunohistochemistry (Paraffin) to compare the spatiotemporal expression of thrombospondin-5 and thrombospondin-4 during postnatal bone formation and fracture healing.
Wed Sep 01 00:00:00 EDT 2021
Tuning the Phenotype of Cartilage Tissue Mimics by Varying Spheroid Maturation and Methacrylamide-Modified Gelatin Hydrogel Characteristics.
Macromolecular bioscience
De Moor L,Minne M,Tytgat L,Vercruysse C,Dubruel P,Van Vlierberghe S,Declercq H
14-9771-82 was used in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) to study how the phenotype of spheroid-laden constructs can be tuned through adjusting the hydrogel physico-chemical properties and if the spheroid maturation stage prior to encapsulation is a determining factor for the construct phenotype.
Sat May 01 00:00:00 EDT 2021
Evidence that TD-198946 enhances the chondrogenic potential of human synovium-derived stem cells through the NOTCH3 signaling pathway.
Journal of tissue engineering and regenerative medicine
Kobayashi M,Chijimatsu R,Hart DA,Hamamoto S,Jacob G,Yano F,Saito T,Shimomura K,Ando W,Chung UI,Tanaka S,Yoshikawa H,Nakamura N
14-9771-82 was used in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) to examine the effect of TD-198946 pretreatment on hSSCs.
Mon Feb 01 00:00:00 EST 2021
Control of skeletal morphogenesis by the Hippo-YAP/TAZ pathway.
Development (Cambridge, England)
Vanyai HK,Prin F,Guillermin O,Marzook B,Boeing S,Howson A,Saunders RE,Snoeks T,Howell M,Mohun TJ,Thompson B
14-9771 was used in Immunohistochemistry to investigate the function of the Hippo pathway during the development of cartilage, which forms the majority of the skeleton.
Thu Nov 12 00:00:00 EST 2020
Lipid phosphatase SHIP-1 regulates chondrocyte hypertrophy and skeletal development.
Journal of cellular physiology
So EY,Sun C,Wu KQ,Driesman A,Leggett S,Isaac M,Spangler T,Dubielecka-Szczerba PM,Reginato AM,Liang OD
Published figure using Collagen X monoclonal antibody (Product # 14-9771-82) in Immunohistochemistry
Sat Feb 01 00:00:00 EST 2020
Angiogenic Potential of Tissue Engineered Cartilage From Human Mesenchymal Stem Cells Is Modulated by Indian Hedgehog and Serpin E1.
Frontiers in bioengineering and biotechnology
Nossin Y,Farrell E,Koevoet WJLM,Somoza RA,Caplan AI,Brachvogel B,van Osch GJVM
14-9771 was used in Immunohistochemistry to identify factors secreted by chondrogenically differentiated bone marrow-derived human BMSCs to modulate angiogenesis.
Thu Dec 31 00:00:00 EST 2020
Angiogenic Potential of Tissue Engineered Cartilage From Human Mesenchymal Stem Cells Is Modulated by Indian Hedgehog and Serpin E1.
Frontiers in bioengineering and biotechnology
Nossin Y,Farrell E,Koevoet WJLM,Somoza RA,Caplan AI,Brachvogel B,van Osch GJVM
14-9771 was used in Immunohistochemistry to identify factors secreted by chondrogenically differentiated bone marrow-derived human BMSCs to modulate angiogenesis.
Thu Dec 31 00:00:00 EST 2020
Impact of the Sensory and Sympathetic Nervous System on Fracture Healing in Ovariectomized Mice.
International journal of molecular sciences
Niedermair T,Straub RH,Brochhausen C,Grässel S
Published figure using Collagen X monoclonal antibody (Product # 14-9771-82) in Immunohistochemistry
Wed Jan 08 00:00:00 EST 2020
Promoting Effect of Basic Fibroblast Growth Factor in Synovial Mesenchymal Stem Cell-Based Cartilage Regeneration.
International journal of molecular sciences
Okamura G,Ebina K,Hirao M,Chijimatsu R,Yonetani Y,Etani Y,Miyama A,Takami K,Goshima A,Yoshikawa H,Ishimoto T,Nakano T,Hamada M,Kanamoto T,Nakata K
14-9771 was used in Immunohistochemistry to investigate the promoting effect of bFGF on cartilage regeneration using human SMSC in vivo.
Wed Dec 30 00:00:00 EST 2020
Promoting Effect of Basic Fibroblast Growth Factor in Synovial Mesenchymal Stem Cell-Based Cartilage Regeneration.
International journal of molecular sciences
Okamura G,Ebina K,Hirao M,Chijimatsu R,Yonetani Y,Etani Y,Miyama A,Takami K,Goshima A,Yoshikawa H,Ishimoto T,Nakano T,Hamada M,Kanamoto T,Nakata K
14-9771 was used in Immunohistochemistry to investigate the promoting effect of bFGF on cartilage regeneration using human SMSC in vivo.
Wed Dec 30 00:00:00 EST 2020
Perlecan Knockdown Significantly Alters Extracellular Matrix Composition and Organization During Cartilage Development.
Molecular & cellular proteomics : MCP
Ocken AR,Ku MM,Kinzer-Ursem TL,Calve S
14-9771-82 was used in Immunohistochemistry to investigate the influence of perlecan on functional cartilage assembly and the progression of osteoarthritis in Schwartz-Jampel syndrome.
Wed Jul 01 00:00:00 EDT 2020
Oral administration of EP4-selective agonist KAG-308 suppresses mouse knee osteoarthritis development through reduction of chondrocyte hypertrophy and TNF secretion.
Scientific reports
Murahashi Y,Yano F,Chijimatsu R,Nakamoto H,Maenohara Y,Amakawa M,Miyake Y,Yamanaka H,Iba K,Yamashita T,Tanaka S,Saito T
14-9771 was used in Immunohistochemistry to examine the efficacy of oral administration of KAG-308, an EP4-selective agonist, in surgically induced mouse knee OA.
Mon Dec 30 00:00:00 EST 2019
Oral administration of EP4-selective agonist KAG-308 suppresses mouse knee osteoarthritis development through reduction of chondrocyte hypertrophy and TNF secretion.
Scientific reports
Murahashi Y,Yano F,Chijimatsu R,Nakamoto H,Maenohara Y,Amakawa M,Miyake Y,Yamanaka H,Iba K,Yamashita T,Tanaka S,Saito T
14-9771 was used in Immunohistochemistry to examine the efficacy of oral administration of KAG-308, an EP4-selective agonist, in surgically induced mouse knee OA.
Mon Dec 30 00:00:00 EST 2019
Oral administration of EP4-selective agonist KAG-308 suppresses mouse knee osteoarthritis development through reduction of chondrocyte hypertrophy and TNF secretion.
Scientific reports
Murahashi Y,Yano F,Chijimatsu R,Nakamoto H,Maenohara Y,Amakawa M,Miyake Y,Yamanaka H,Iba K,Yamashita T,Tanaka S,Saito T
14-9771 was used in Immunohistochemistry to examine the efficacy of oral administration of KAG-308, an EP4-selective agonist, in surgically induced mouse knee OA.
Mon Dec 30 00:00:00 EST 2019
Phenotypic Characterization of Bone Marrow Mononuclear Cells and Derived Stromal Cell Populations from Human Iliac Crest, Vertebral Body and Femoral Head.
International journal of molecular sciences
Herrmann M,Hildebrand M,Menzel U,Fahy N,Alini M,Lang S,Benneker L,Verrier S,Stoddart MJ,Bara JJ
14-9771 was used in Immunohistochemistry to suggest more reliable sources of tissue of stromal progenitor cells for the repair of cartilage.
Sun Jul 14 00:00:00 EDT 2019
A Hydrogel Model Incorporating 3D-Plotted Hydroxyapatite for Osteochondral Tissue Engineering.
Materials (Basel, Switzerland)
Bartnikowski M,Akkineni AR,Gelinsky M,Woodruff MA,Klein TJ
Published figure using Collagen X monoclonal antibody (Product # 14-9771-82) in Immunofluorescence
Thu Apr 14 00:00:00 EDT 2016
A Hydrogel Model Incorporating 3D-Plotted Hydroxyapatite for Osteochondral Tissue Engineering.
Materials (Basel, Switzerland)
Bartnikowski M,Akkineni AR,Gelinsky M,Woodruff MA,Klein TJ
Published figure using Collagen X monoclonal antibody (Product # 14-9771-82) in Immunofluorescence
Thu Apr 14 00:00:00 EDT 2016
Identification of stromal ColXα1 and tumor-infiltrating lymphocytes as putative predictive markers of neoadjuvant therapy in estrogen receptor-positive/HER2-positive breast cancer.
BMC cancer
Brodsky AS,Xiong J,Yang D,Schorl C,Fenton MA,Graves TA,Sikov WM,Resnick MB,Wang Y
14-9771 was used in Immunohistochemistry to monitor protein expression during neoadjuvant therapy for breast cancer and compare expression to pathologic response.
Mon Apr 18 00:00:00 EDT 2016
The gonadotrophin-releasing hormone receptor of alpha T3-1 pituitary cells regulates cellular levels of both of the phosphoinositidase C-linked G proteins, Gq alpha and G11 alpha, equally.
Molecular pharmacology
Shah BH,Milligan G
Published figure using Collagen X monoclonal antibody (Product # 14-9771-82) in Western Blot
Fri Jul 01 00:00:00 EDT 1994
The gonadotrophin-releasing hormone receptor of alpha T3-1 pituitary cells regulates cellular levels of both of the phosphoinositidase C-linked G proteins, Gq alpha and G11 alpha, equally.
Molecular pharmacology
Shah BH,Milligan G
Published figure using Collagen X monoclonal antibody (Product # 14-9771-82) in Western Blot
Fri Jul 01 00:00:00 EDT 1994