Product References
Significance of Plasmodium berghei Amino Acid Transporter 1 in Food Vacuole Functionality and Its Association with Cerebral Pathogenesis.
Microbiology spectrum
Anand A,Chandana M,Ghosh S,Das R,Singh N,Vaishalli PM,Gantasala NP,Padmanaban G,Nagaraj VA
14-9832-82 was used in Western Blotting to show that the deletion of amino acid transporter 1 in Plasmodium berghei leads to swollen food vacuoles with the accumulation of hemoglobin-derived peptides.
Tue Mar 28 00:00:00 EDT 2023
The p-STAT3/ANXA2 axis promotes caspase-1-mediated hepatocyte pyroptosis in non-alcoholic steatohepatitis.
Journal of translational medicine
Feng Y,Li W,Wang Z,Zhang R,Li Y,Zang L,Wang P,Li Z,Dong Y
Published figure using Caspase 1 monoclonal antibody (Product # 14-9832-82) in Western Blot
Wed Nov 02 00:00:00 EDT 2022
Glycosylation-Dependent Induction of Programmed Cell Death in Murine Adenocarcinoma Cells.
Frontiers in immunology
Parshenkov A,Hennet T
Published figure using Caspase 1 monoclonal antibody (Product # 14-9832-82) in Western Blot
Fri Apr 08 00:00:00 EDT 2022
Caspase-11 interaction with NLRP3 potentiates the noncanonical activation of the NLRP3 inflammasome.
Nature immunology
Moretti J,Jia B,Hutchins Z,Roy S,Yip H,Wu J,Shan M,Jaffrey SR,Coers J,Blander JM
14-9832-82 was used in Western Blotting to find that bacterial mRNA, which signals bacterial viability, was required alongside LPS for noncanonical activation of the NLRP3 inflammasome in macrophages.
Sun May 01 00:00:00 EDT 2022
Adipocyte CD1d Gene Transfer Induces T Cell Expansion and Adipocyte Inflammation in CD1d Knockout Mice.
Journal of immunology (Baltimore, Md. : 1950)
Xiao R,Mansour AG,Huang W,Hassan QN,Wilkins RK,Komatineni SV,Bates R,Ali S,Chrislip LA,Queen NJ,Ma S,Yu J,Lordo MR,Mundy-Bosse BL,Caligiuri MA,Cao L
14-9832-82 was used in Western Blotting to report that adeno-associated virus-mediated gene transfer of CD1d to adipocytes in CD1d KO mice fails to rescue iNKT cells but leads to massive and selective expansion of T cells within adipose tissue, particularly CD8+ T effector cells, that is associated with adipocyte NLRP3 inflammasome activation, dysregulation of adipocyte functional genes, and upregulation of apoptotic pathway proteins.
Sun May 01 00:00:00 EDT 2022
Malaria parasite heme biosynthesis promotes and griseofulvin protects against cerebral malaria in mice.
Nature communications
Chandana M,Anand A,Ghosh S,Das R,Beura S,Jena S,Suryawanshi AR,Padmanaban G,Nagaraj VA
14-9832-82 was used in Western Blotting to show heme synthesized in asexual stages promotes cerebral pathogenesis by enhancing hemozoin formation.
Tue Jul 12 00:00:00 EDT 2022
Modulation of the NLRP3 inflammasome by Sars-CoV-2 Envelope protein.
Scientific reports
Yalcinkaya M,Liu W,Islam MN,Kotini AG,Gusarova GA,Fidler TP,Papapetrou EP,Bhattacharya J,Wang N,Tall AR
Published figure using Caspase 1 monoclonal antibody (Product # 14-9832-82) in Western Blot
Fri Dec 24 00:00:00 EST 2021
Integrative metabolomics and transcriptomics identifies itaconate as an adjunct therapy to treat ocular bacterial infection.
Cell reports. Medicine
Singh S,Singh PK,Jha A,Naik P,Joseph J,Giri S,Kumar A
14-9832-82 was used in Western Blot to study the anti-inflammatory effects of itaconate using integrated metabolomics and transcriptomics.
Tue May 18 00:00:00 EDT 2021
NLPR3 inflammasome inhibition alleviates hypoxic endothelial cell death in vitro and protects blood-brain barrier integrity in murine stroke.
Cell death & disease
Bellut M,Papp L,Bieber M,Kraft P,Stoll G,Schuhmann MK
14-9832-82 was used in Immunohistochemistry (Frozen) to study the survival of brain endothelial cells exposed to oxygen and glucose deprivation in vitro in the context of inflammasome inhibition with the NLR-family pyrin domain-containing protein 3 (NLRP3)-specific drug MCC950.
Mon Dec 20 00:00:00 EST 2021
Caspase-8-Dependent Inflammatory Responses Are Controlled by Its Adaptor, FADD, and Necroptosis.
Immunity
Tummers B,Mari L,Guy CS,Heckmann BL,Rodriguez DA,Rühl S,Moretti J,Crawford JC,Fitzgerald P,Kanneganti TD,Janke LJ,Pelletier S,Blander JM,Green DR
14-9832-82 was used in Western Blot to suggest an in vivo role of the inflammatory RIPK1-caspase-8-FADD (FADDosome) complex and reveal a FADD-independent inflammatory role of caspase-8 that involves activation of an inflammasome.
Tue Jun 16 00:00:00 EDT 2020
An Acetylation Switch of the NLRP3 Inflammasome Regulates Aging-Associated Chronic Inflammation and Insulin Resistance.
Cell metabolism
He M,Chiang HH,Luo H,Zheng Z,Qiao Q,Wang L,Tan M,Ohkubo R,Mu WC,Zhao S,Wu H,Chen D
14-9832 was used in Western Blotting to establish the dysregulation of the acetylation switch of the NLRP3 inflammasome as an origin of aging-associated chronic inflammation and highlight the reversibility of aging-associated chronic inflammation and insulin resistance.
Tue Mar 03 00:00:00 EST 2020
Long non-coding RNA Gm15441 attenuates hepatic inflammasome activation in response to PPARA agonism and fasting.
Nature communications
Brocker CN,Kim D,Melia T,Karri K,Velenosi TJ,Takahashi S,Aibara D,Bonzo JA,Levi M,Waxman DJ,Gonzalez FJ
14-9832 was used in Western Blotting to show nuclear receptor PPARα reduces inflammasome activation through induction of long non-coding RNA Gm15441 as a result of metabolic stress.
Tue Nov 17 00:00:00 EST 2020
Long non-coding RNA Gm15441 attenuates hepatic inflammasome activation in response to PPARA agonism and fasting.
Nature communications
Brocker CN,Kim D,Melia T,Karri K,Velenosi TJ,Takahashi S,Aibara D,Bonzo JA,Levi M,Waxman DJ,Gonzalez FJ
14-9832 was used in Western Blotting to show nuclear receptor PPARα reduces inflammasome activation through induction of long non-coding RNA Gm15441 as a result of metabolic stress.
Tue Nov 17 00:00:00 EST 2020
Cutting Edge: TAK1 Safeguards Macrophages against Proinflammatory Cell Death.
Journal of immunology (Baltimore, Md. : 1950)
Sanjo H,Nakayama J,Yoshizawa T,Fehling HJ,Akira S,Taki S
14-9832-82 was used in Cell Culture to reveal that TAK1 acts as a signaling checkpoint to protect macrophages from unique proinflammatory cell death, ensuring the maintenance of innate immune homeostasis.
Thu Aug 15 00:00:00 EDT 2019
Mitochondrial Stress-Initiated Aberrant Activation of the NLRP3 Inflammasome Regulates the Functional Deterioration of Hematopoietic Stem Cell Aging.
Cell reports
Luo H,Mu WC,Karki R,Chiang HH,Mohrin M,Shin JJ,Ohkubo R,Ito K,Kanneganti TD,Chen D
14-9832 was used in Western Blotting to assess whether the NLRP3 inflammasome is aberrantly activated in haematopoietic stem cells during physiological ageing.
Tue Jan 22 00:00:00 EST 2019
Mitochondrial Stress-Initiated Aberrant Activation of the NLRP3 Inflammasome Regulates the Functional Deterioration of Hematopoietic Stem Cell Aging.
Cell reports
Luo H,Mu WC,Karki R,Chiang HH,Mohrin M,Shin JJ,Ohkubo R,Ito K,Kanneganti TD,Chen D
14-9832 was used in Western Blotting to assess whether the NLRP3 inflammasome is aberrantly activated in haematopoietic stem cells during physiological ageing.
Tue Jan 22 00:00:00 EST 2019
Intra-amniotic inflammation induces preterm birth by activating the NLRP3 inflammasome†.
Biology of reproduction
Faro J,Romero R,Schwenkel G,Garcia-Flores V,Arenas-Hernandez M,Leng Y,Xu Y,Miller D,Hassan SS,Gomez-Lopez N
14-9832-82 was used in Western Blotting to investigate effects of NLRP3 inflammosome induction on preterm birth.
Wed May 01 00:00:00 EDT 2019
Free immunoglobulin light chain (FLC) promotes murine colitis and colitis-associated colon carcinogenesis by activating the inflammasome.
Scientific reports
Ma J,Jiang D,Gong X,Shao W,Zhu Z,Xu W,Qiu X
14-9832 was used in Western Blotting to investigate whether free Ig light chain is involved in colitis and colitis-associated colon carcinogenesis.
Wed Jul 12 00:00:00 EDT 2017
Neutrophil extracellular traps enriched in oxidized mitochondrial DNA are interferogenic and contribute to lupus-like disease.
Nature medicine
Lood C,Blanco LP,Purmalek MM,Carmona-Rivera C,De Ravin SS,Smith CK,Malech HL,Ledbetter JA,Elkon KB,Kaplan MJ
14-9832 was used in Western Blotting to investigate how neutrophil extracellular traps are generated and their roles in sterile inflammation.
Mon Feb 01 00:00:00 EST 2016
Identification of QS-21 as an Inflammasome-activating Molecular Component of Saponin Adjuvants.
The Journal of biological chemistry
Marty-Roix R,Vladimer GI,Pouliot K,Weng D,Buglione-Corbett R,West K,MacMicking JD,Chee JD,Wang S,Lu S,Lien E
14-9832 was used in Western Blotting to determine the mechanisms of cellular action induced by QS-21.
Fri Jan 15 00:00:00 EST 2016
Bruton's tyrosine kinase is essential for NLRP3 inflammasome activation and contributes to ischaemic brain injury.
Nature communications
Ito M,Shichita T,Okada M,Komine R,Noguchi Y,Yoshimura A,Morita R
14-9832 was used in Immunoprecipitation to report that Bruton's tyrosine kinase (BTK) is an essential component of the NLRP3 inflammasome, in which BTK physically interacts with ASC and NLRP3.
Wed Jun 10 00:00:00 EDT 2015
Nitric oxide controls the immunopathology of tuberculosis by inhibiting NLRP3 inflammasome-dependent processing of IL-1β.
Nature immunology
Mishra BB,Rathinam VA,Martens GW,Martinot AJ,Kornfeld H,Fitzgerald KA,Sassetti CM
14-9832 was used in Western Blotting to investigate the mechanisms that controls IL-1 during tuberculosis.
Tue Jan 01 00:00:00 EST 2013
Synthetic oligodeoxynucleotides containing suppressive TTAGGG motifs inhibit AIM2 inflammasome activation.
Journal of immunology (Baltimore, Md. : 1950)
Kaminski JJ,Schattgen SA,Tzeng TC,Bode C,Klinman DM,Fitzgerald KA
14-9832 was used in Western Blotting to investigate the effect of ODN-A151 in dendritic cells and macrophages.
Tue Oct 01 00:00:00 EDT 2013
TRIF licenses caspase-11-dependent NLRP3 inflammasome activation by gram-negative bacteria.
Cell
Rathinam VA,Vanaja SK,Waggoner L,Sokolovska A,Becker C,Stuart LM,Leong JM,Fitzgerald KA
14-9832 was used in Western Blotting to investigate the molecular mechanisms underlying "sepsis syndrome" response during systemic Gram-negative bacteria infection.
Fri Aug 03 00:00:00 EDT 2012
Chitosan but not chitin activates the inflammasome by a mechanism dependent upon phagocytosis.
The Journal of biological chemistry
Bueter CL,Lee CK,Rathinam VAK,Healy GJ,Taron CH,Specht CA,Levitz SM
14-9832 was used in Western Blotting to investigate the capacity of chitin to stimulate murine macrophages to release the inflammasome-associated cytokine IL-1β.
Fri Oct 14 00:00:00 EDT 2011