Product References
Label-Free Proteomic Analysis of Exosomes Secreted from THP-1-Derived Macrophages Treated with IFN-α Identifies Antiviral Proteins Enriched in Exosomes.
Journal of proteome research
Yao Z,Jia X,Megger DA,Chen J,Liu Y,Li J,Sitek B,Yuan Z
Fri Mar 01 00:00:00 EST 2019
mTOR inhibitors lower an intrinsic barrier to virus infection mediated by IFITM3.
Proceedings of the National Academy of Sciences of the United States of America
Shi G,Ozog S,Torbett BE,Compton AA
Tue Oct 23 00:00:00 EDT 2018
Exosomes From Adipose-Derived Stem Cells Attenuate Adipose Inflammation and Obesity Through Polarizing M2 Macrophages and Beiging in White Adipose Tissue.
Diabetes
Zhao H,Shang Q,Pan Z,Bai Y,Li Z,Zhang H,Zhang Q,Guo C,Zhang L,Wang Q
Thu Feb 01 00:00:00 EST 2018
Circulating Exosomal miR-17-5p and miR-92a-3p Predict Pathologic Stage and Grade of Colorectal Cancer.
Translational oncology
Fu F,Jiang W,Zhou L,Chen Z
Sun Apr 01 00:00:00 EDT 2018
Adipose mesenchymal stem cell-derived exosomes stimulated by hydrogen peroxide enhanced skin flap recovery in ischemia-reperfusion injury.
Biochemical and biophysical research communications
Bai Y,Han YD,Yan XL,Ren J,Zeng Q,Li XD,Pei XT,Han Y
Sat Jun 02 00:00:00 EDT 2018
Exosomes derived from breast cancer lung metastasis subpopulations promote tumor self-seeding.
Biochemical and biophysical research communications
Huang H,Zheng X,Cai C,Yao Z,Lu S,Meng X,Miao Y,He Z,Cai C,Zou F
Mon Sep 03 00:00:00 EDT 2018
Co-transplantation of exosomes derived from hypoxia-preconditioned adipose mesenchymal stem cells promotes neovascularization and graft survival in fat grafting.
Biochemical and biophysical research communications
Han YD,Bai Y,Yan XL,Ren J,Zeng Q,Li XD,Pei XT,Han Y
Mon Feb 26 00:00:00 EST 2018
Exosomes from human umbilical cord blood accelerate cutaneous wound healing through miR-21-3p-mediated promotion of angiogenesis and fibroblast function.
Theranostics
Hu Y,Rao SS,Wang ZX,Cao J,Tan YJ,Luo J,Li HM,Zhang WS,Chen CY,Xie H
Mon Nov 19 00:00:00 EST 2018
Melanoma cell-secreted exosomal miR-155-5p induce proangiogenic switch of cancer-associated fibroblasts via SOCS1/JAK2/STAT3 signaling pathway.
Journal of experimental & clinical cancer research : CR
Zhou X,Yan T,Huang C,Xu Z,Wang L,Jiang E,Wang H,Chen Y,Liu K,Shao Z,Shang Z
Wed Oct 03 00:00:00 EDT 2018
Exosomal DMBT1 from human urine-derived stem cells facilitates diabetic wound repair by promoting angiogenesis.
Theranostics
Chen CY,Rao SS,Ren L,Hu XK,Tan YJ,Hu Y,Luo J,Liu YW,Yin H,Huang J,Cao J,Wang ZX,Liu ZZ,Liu HM,Tang SY,Xu R,Xie H
Wed Jan 30 00:00:00 EST 2019
Characterization and selective incorporation of small non-coding RNAs in non-small cell lung cancer extracellular vesicles.
Cell & bioscience
Li C,Qin F,Hu F,Xu H,Sun G,Han G,Wang T,Guo M
Fri Mar 18 00:00:00 EDT 2022
Tumor-Derived Exosomal Long Noncoding RNAs as Promising Diagnostic Biomarkers for Prostate Cancer.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang YH,Ji J,Wang BC,Chen H,Yang ZH,Wang K,Luo CL,Zhang WW,Wang FB,Zhang XL
Thu Jul 05 00:00:00 EDT 2018
Exosome RNA Unshielding Couples Stromal Activation to Pattern Recognition Receptor Signaling in Cancer.
Cell
Nabet BY,Qiu Y,Shabason JE,Wu TJ,Yoon T,Kim BC,Benci JL,DeMichele AM,Tchou J,Marcotrigiano J,Minn AJ
Thu Jul 13 00:00:00 EDT 2017
Exosome-mediated miR-146a transfer suppresses type I interferon response and facilitates EV71 infection.
PLoS pathogens
Fu Y,Zhang L,Zhang F,Tang T,Zhou Q,Feng C,Jin Y,Wu Z
Fri Sep 01 00:00:00 EDT 2017
REG3β modifies cell tumor function by impairing extracellular vesicle uptake.
Scientific reports
Bonjoch L,Gironella M,Iovanna JL,Closa D
Fri Jun 09 00:00:00 EDT 2017
Exosomes maintain cellular homeostasis by excreting harmful DNA from cells.
Nature communications
Takahashi A,Okada R,Nagao K,Kawamata Y,Hanyu A,Yoshimoto S,Takasugi M,Watanabe S,Kanemaki MT,Obuse C,Hara E
Tue May 16 00:00:00 EDT 2017
Label-free Proteomic Analysis of Exosomes Derived from Inducible Hepatitis B Virus-Replicating HepAD38 Cell Line.
Molecular & cellular proteomics : MCP
Jia X,Chen J,Megger DA,Zhang X,Kozlowski M,Zhang L,Fang Z,Li J,Chu Q,Wu M,Li Y,Sitek B,Yuan Z
Sat Apr 01 00:00:00 EDT 2017
Charcot-Marie-Tooth disease type 2G redefined by a novel mutation in LRSAM1.
Annals of neurology
Peeters K,Palaima P,Pelayo-Negro AL,García A,Gallardo E,García-Barredo R,Mateiu L,Baets J,Menten B,De Vriendt E,De Jonghe P,Timmerman V,Infante J,Berciano J,Jordanova A
Thu Dec 01 00:00:00 EST 2016
Involvement of exosomes in lung inflammation associated with experimental acute pancreatitis.
The Journal of pathology
Bonjoch L,Casas V,Carrascal M,Closa D
Sat Oct 01 00:00:00 EDT 2016
ESCRT-I Protein Tsg101 Plays a Role in the Post-macropinocytic Trafficking and Infection of Endothelial Cells by Kaposi's Sarcoma-Associated Herpesvirus.
PLoS pathogens
Kumar B,Dutta D,Iqbal J,Ansari MA,Roy A,Chikoti L,Pisano G,Veettil MV,Chandran B
Sat Oct 01 00:00:00 EDT 2016
ESCRT-I Protein Tsg101 Plays a Role in the Post-macropinocytic Trafficking and Infection of Endothelial Cells by Kaposi's Sarcoma-Associated Herpesvirus.
PLoS pathogens
Kumar B,Dutta D,Iqbal J,Ansari MA,Roy A,Chikoti L,Pisano G,Veettil MV,Chandran B
Sat Oct 01 00:00:00 EDT 2016
Oligodendroglial deletion of ESCRT-I component TSG101 causes spongiform encephalopathy.
Biology of the cell
Walker WP,Oehler A,Edinger AL,Wagner KU,Gunn TM
Tue Nov 01 00:00:00 EDT 2016
Characterization of exosomes derived from ovarian cancer cells and normal ovarian epithelial cells by nanoparticle tracking analysis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Zhang W,Peng P,Kuang Y,Yang J,Cao D,You Y,Shen K
Tue Mar 01 00:00:00 EST 2016
Regulation of exosomes released from normal ovarian epithelial cells and ovarian cancer cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Zhang W,Yang J,Cao D,You Y,Shen K,Peng P
Thu Oct 06 00:00:00 EDT 2016
Exosomes play an important role in the process of psoralen reverse multidrug resistance of breast cancer.
Journal of experimental & clinical cancer research : CR
Wang X,Xu C,Hua Y,Sun L,Cheng K,Jia Z,Han Y,Dong J,Cui Y,Yang Z
Thu Dec 01 00:00:00 EST 2016
Microenvironment-induced PTEN loss by exosomal microRNA primes brain metastasis outgrowth.
Nature
Zhang L,Zhang S,Yao J,Lowery FJ,Zhang Q,Huang WC,Li P,Li M,Wang X,Zhang C,Wang H,Ellis K,Cheerathodi M,McCarty JH,Palmieri D,Saunus J,Lakhani S,Huang S,Sahin AA,Aldape KD,Steeg PS,Yu D
Thu Nov 05 00:00:00 EST 2015
Microenvironment-induced PTEN loss by exosomal microRNA primes brain metastasis outgrowth.
Nature
Zhang L,Zhang S,Yao J,Lowery FJ,Zhang Q,Huang WC,Li P,Li M,Wang X,Zhang C,Wang H,Ellis K,Cheerathodi M,McCarty JH,Palmieri D,Saunus J,Lakhani S,Huang S,Sahin AA,Aldape KD,Steeg PS,Yu D
Thu Nov 05 00:00:00 EST 2015
High LIN28A Expressing Ovarian Cancer Cells Secrete Exosomes That Induce Invasion and Migration in HEK293 Cells.
BioMed research international
Enriquez VA,Cleys ER,Da Silveira JC,Spillman MA,Winger QA,Bouma GJ
Tue Sep 06 00:00:00 EDT 2016
Exosome transfer from stromal to breast cancer cells regulates therapy resistance pathways.
Cell
Boelens MC,Wu TJ,Nabet BY,Xu B,Qiu Y,Yoon T,Azzam DJ,Twyman-Saint Victor C,Wiemann BZ,Ishwaran H,Ter Brugge PJ,Jonkers J,Slingerland J,Minn AJ
Thu Oct 23 00:00:00 EDT 2014
Exosomes mediate the cell-to-cell transmission of IFN-α-induced antiviral activity.
Nature immunology
Li J,Liu K,Liu Y,Xu Y,Zhang F,Yang H,Liu J,Pan T,Chen J,Wu M,Zhou X,Yuan Z
Thu Aug 01 00:00:00 EDT 2013
Characterization and proteomic analysis of ovarian cancer-derived exosomes.
Journal of proteomics
Liang B,Peng P,Chen S,Li L,Zhang M,Cao D,Yang J,Li H,Gui T,Li X,Shen K
Wed Mar 27 00:00:00 EDT 2013
Molecular crosstalk between cancer cells and tumor microenvironment components suggests potential targets for new therapeutic approaches in mobile tongue cancer.
Cancer medicine
Dayan D,Salo T,Salo S,Nyberg P,Nurmenniemi S,Costea DE,Vered M
Mon Oct 01 00:00:00 EDT 2012