Product References
NLRP3 Inflammasome Activation of Mast Cells by Estrogen via the Nuclear-Initiated Signaling Pathway Contributes to the Development of Endometriosis.
Frontiers in immunology
Guo X,Xu X,Li T,Yu Q,Wang J,Chen Y,Ding S,Zhu L,Zou G,Zhang X
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Tue Jan 18 00:00:00 EST 2022
SNAP23 is essential for platelet and mast cell development and required in connective tissue mast cells for anaphylaxis.
The Journal of biological chemistry
Cardenas RA,Gonzalez R,Sanchez E,Ramos MA,Cardenas EI,Rodarte AI,Alcazar-Felix RJ,Isaza A,Burns AR,Heidelberger R,Adachi R
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
Mutant Huntingtin affects toll-like receptor 4 intracellular trafficking and cytokine production in mast cells.
Journal of neuroinflammation
Pérez-Rodríguez MJ,Ibarra-Sánchez A,Román-Figueroa A,Pérez-Severiano F,González-Espinosa C
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Fri Mar 27 00:00:00 EDT 2020
Tespa1 plays a role in the modulation of airway hyperreactivity through the IL-4/STAT6 pathway.
Journal of translational medicine
Yang R,Wang G,Li L,He H,Zheng M,Lu L,Wu S
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Mon Nov 23 00:00:00 EST 2020
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
Type I interferon limits mast cell-mediated anaphylaxis by controlling secretory granule homeostasis.
PLoS biology
Kobayashi T,Shimabukuro-Demoto S,Tsutsui H,Toyama-Sorimachi N
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2019
Na+-H+ exchanger 1 determines atherosclerotic lesion acidification and promotes atherogenesis.
Nature communications
Liu CL,Zhang X,Liu J,Wang Y,Sukhova GK,Wojtkiewicz GR,Liu T,Tang R,Achilefu S,Nahrendorf M,Libby P,Guo J,Zhang JY,Shi GP
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Western Blot
Wed Sep 04 00:00:00 EDT 2019
Positive and Negative Regulatory Roles of C-Terminal Src Kinase (CSK) in FcεRI-Mediated Mast Cell Activation, Independent of the Transmembrane Adaptor PAG/CSK-Binding Protein.
Frontiers in immunology
Potuckova L,Draberova L,Halova I,Paulenda T,Draber P
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Mon Oct 07 00:00:00 EDT 2019
Positive and Negative Regulatory Roles of C-Terminal Src Kinase (CSK) in FcεRI-Mediated Mast Cell Activation, Independent of the Transmembrane Adaptor PAG/CSK-Binding Protein.
Frontiers in immunology
Potuckova L,Draberova L,Halova I,Paulenda T,Draber P
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Mon Oct 07 00:00:00 EDT 2019
Positive and Negative Regulatory Roles of C-Terminal Src Kinase (CSK) in FcεRI-Mediated Mast Cell Activation, Independent of the Transmembrane Adaptor PAG/CSK-Binding Protein.
Frontiers in immunology
Potuckova L,Draberova L,Halova I,Paulenda T,Draber P
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Mon Oct 07 00:00:00 EDT 2019
Mast cell-derived neurotrophin 4 mediates allergen-induced airway hyperinnervation in early life.
Mucosal immunology
Patel KR,Aven L,Shao F,Krishnamoorthy N,Duvall MG,Levy BD,Ai X
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2016
New Regulatory Roles of Galectin-3 in High-Affinity IgE Receptor Signaling.
Molecular and cellular biology
Bambouskova M,Polakovicova I,Halova I,Goel G,Draberova L,Bugajev V,Doan A,Utekal P,Gardet A,Xavier RJ,Draber P
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
New Regulatory Roles of Galectin-3 in High-Affinity IgE Receptor Signaling.
Molecular and cellular biology
Bambouskova M,Polakovicova I,Halova I,Goel G,Draberova L,Bugajev V,Doan A,Utekal P,Gardet A,Xavier RJ,Draber P
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
Isolation of Mature (Peritoneum-Derived) Mast Cells and Immature (Bone Marrow-Derived) Mast Cell Precursors from Mice.
PloS one
Meurer SK,Neß M,Weiskirchen S,Kim P,Tag CG,Kauffmann M,Huber M,Weiskirchen R
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Mon Jul 24 00:00:00 EDT 2017
Deficiency of FcϵR1 Increases Body Weight Gain but Improves Glucose Tolerance in Diet-Induced Obese Mice.
Endocrinology
Lee YJ,Liu C,Liao M,Sukhova GK,Shirakawa J,Abdennour M,Iamarene K,Andre S,Inouye K,Clement K,Kulkarni RN,Banks AS,Libby P,Shi GP
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Western Blot
Sun Nov 01 00:00:00 EDT 2015
A next-generation dual-recombinase system for time- and host-specific targeting of pancreatic cancer.
Nature medicine
Schönhuber N,Seidler B,Schuck K,Veltkamp C,Schachtler C,Zukowska M,Eser S,Feyerabend TB,Paul MC,Eser P,Klein S,Lowy AM,Banerjee R,Yang F,Lee CL,Moding EJ,Kirsch DG,Scheideler A,Alessi DR,Varela I,Bradley A,Kind A,Schnieke AE,Rodewald HR,Rad R,Schmid RM,Schneider G,Saur D
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Immunofluorescence
Sat Nov 01 00:00:00 EDT 2014
MHC class II-dependent basophil-CD4+ T cell interactions promote T(H)2 cytokine-dependent immunity.
Nature immunology
Perrigoue JG,Saenz SA,Siracusa MC,Allenspach EJ,Taylor BC,Giacomin PR,Nair MG,Du Y,Zaph C,van Rooijen N,Comeau MR,Pearce EJ,Laufer TM,Artis D
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2009
MHC class II-dependent basophil-CD4+ T cell interactions promote T(H)2 cytokine-dependent immunity.
Nature immunology
Perrigoue JG,Saenz SA,Siracusa MC,Allenspach EJ,Taylor BC,Giacomin PR,Nair MG,Du Y,Zaph C,van Rooijen N,Comeau MR,Pearce EJ,Laufer TM,Artis D
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2009
IL-21R is essential for epicutaneous sensitization and allergic skin inflammation in humans and mice.
The Journal of clinical investigation
Jin H,Oyoshi MK,Le Y,Bianchi T,Koduru S,Mathias CB,Kumar L,Le Bras S,Young D,Collins M,Grusby MJ,Wenzel J,Bieber T,Boes M,Silberstein LE,Oettgen HC,Geha RS
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2009
Mast cell degranulation breaks peripheral tolerance.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
de Vries VC,Wasiuk A,Bennett KA,Benson MJ,Elgueta R,Waldschmidt TJ,Noelle RJ
16-5898 was used in Flow cytometry/Cell sorting to show that intragraft or systemic mast cell degranulation results in the transient loss of Treg suppressor activities and the rejection of established allografts.
Thu Oct 01 00:00:00 EDT 2009
Developmental checkpoints of the basophil/mast cell lineages in adult murine hematopoiesis.
Proceedings of the National Academy of Sciences of the United States of America
Arinobu Y,Iwasaki H,Gurish MF,Mizuno S,Shigematsu H,Ozawa H,Tenen DG,Austen KF,Akashi K
Published figure using FceR1 alpha monoclonal antibody (Product # 16-5898-82) in Flow Cytometry
Tue Dec 13 00:00:00 EST 2005