Search Thermo Fisher Scientific
Immunogen sequence: MAEGEDMQT FTSIMDALVR ISTSMKNMEK ELLCPVCQEM YKQPLVLPCT HNVCQACARE VLGQQGYIGH GGDPSSEPTS PASTPSTRSP RLSRRTLPKP DRLDRLLKSG FGTYPGRKRG ALHPQVIMFP CPACQGDVEL GERGLAGLFR NLTLERVVER YRQSVSVGGA ILCQLCKPPP LEATKGCTEC RATFCNECFK LFHPWGTQKA QHEPTLPTLS FRPKGLMCPD HKEEVTHYCK TCQRLVCQLC RVRRTHSGHK ITPVLSAYQA LKDKLTKSLT YILGNQDTVQ TQICELEEAV RHTEVSGQQA KEEVSQLVRG LGAVLEEKRA SLLQAIEECQ QERLARLSAQ I (1-350 aa encoded by BC069416)
Microtubule-associated protein that is involved in the formation of parallel microtubule bundles linked by cross-bridges in the proximal axon. Required for the uniform orientation and maintenance of the parallel microtubule fascicles, which are important for efficient cargo delivery and trafficking in axons. Thereby also required for proper axon specification, the establishment of neuronal polarity and proper neuronal migration. [UniProt]