Search Thermo Fisher Scientific
Immunogen sequence: QQLAAVRGG GGMALPQPQP QSLPLPPSLP SALAPVPSLP SAPAPVPALP LPRRPSALPP KENGCAVWST EATFRPGPEP PPPPPRRRAS APHGPLDMAP FARLHECYGQ EIRALRGQWL SSRVQP (125-249 aa encoded by BC034809)
Plays a role in endocytic trafficking. Required for receptor recycling from endosomes, both to the trans-Golgi network and the plasma membrane.