A2B5 binds to neural tissue such as retina, brain, spinal cord, and dorsal root ganglia. It is also able to bind plasma membranes of human, rat, chicken and mouse islet cells and can be used in cell sorting.
Protein Aliases: A2B5 marker