Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.