Based on 100% sequence identity, this antibody is predicted to react with Bovine, Pig, Orangutan, Monkey and Chimpanzee
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.