Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Transcriptional activator. Binds specifically to the DNA sequence 5'-AACAAT-3'. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.