Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Rabbit, Golden hamster, Orangutan, Monkey, Gorilla and Chimpanzee
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.