Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
beta 3 Adrenergic Receptor is located mainly in adipose tissue and is involved in the regulation of lipolysis and thermogenesis. Beta adrenergic receptors are involved in the epinephrine and norepinephrine-induced activation of adenylate cyclase through the action of G proteins. beta 3 Adrenergic Receptor expression has been documented in adipose tissues, heart, and in smooth muscle of digestive and urinary tract organs (bladder, colon, small intestine, stomach, ureter). Utilization of alternate promoters and/or 3-prime untranslated regions may result in tissue-specific regulation of the expression of this protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenergic, beta-3-, receptor; beta 3 adrenoceptor; Beta-3 adrenergic receptor; Beta-3 adrenoceptor; Beta-3 adrenoreceptor; beta3-Adrenoreceptor; Beta3-AR; FLJ99960
Gene Aliases: ADRB3; ADRB3R; B3AR; BETA3AR
UniProt ID: (Human) P13945
Entrez Gene ID: (Human) 155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support