Search Thermo Fisher Scientific
Immunogen sequence: SIQAANLEE DKLPEIPERG IRTGAAATPE EFEAIAELKS DDWSYLVPMT ELKPGNPASG HNYFLVNSQQ RWTHIRLNIF PDGGIARLRV FGTGQKDWTA TDPKEPADLV AIAFGGVCVG FSNAKFGHPN NIIGVGGAKS MADGWETARR LDRPPILEND ENGILLVPGC EWAVFRLAHP GVITRIEIDT KYFEGNAPDS CKVDGCILTT QEEAVIRQKW ILPAHKWKPL LPVTKLSPNQ SHLFDSLTLE LQDVITHARL TIVPDGGVSR LRLRGFPSSI CLLRPREKPM LKFSVSFKAN P (110-409 aa encoded by BC029652)
Allantoicase (EC 3.5.3.4) participates in the uric acid degradation pathway. Its enzymatic activity, like that of urate oxidase, was lost during vertebrate evolution.