Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: ERDSTGIVTP SGTFHQHVPA GYMDFPKVGG MPFSVYGNAM IPPVAPIPDG AGGPIFNGPH AADPSWNSLI KMVSSSTENN GPQTVWTGPW APHMNSVHMN QLG
This gene encodes a protein with ankyrin repeats, which are associated with protein-protein interactions. Studies in mice suggest that this protein is involved in liver development. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 17; Gene trap ankyrin repeat protein; Serologically defined breast cancer antigen NY-BR-16
Gene Aliases: ANKRD17; GTAR; KIAA0697; MASK2; NY-BR-16
UniProt ID: (Human) O75179
Entrez Gene ID: (Human) 26057
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support