Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Recommended positive controls: 3xFlag-human ARID5A-transfected 293T, THP-1, THP-1 nuclear extract, Raw264.7, Raw264.7 (50 ng/mL LPS, 4hr).
Predicted reactivity: Mouse (83%), Bovine (88%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARID domain-containing protein 5A; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interactive domain-containing protein 5A; Modulator recognition factor 1; modulator recognition factor I; MRF-1; RFVG5814
Gene Aliases: ARID5A; D430024K22Rik; MRF-1; MRF1; RP11-363D14
UniProt ID: (Human) Q03989, (Mouse) Q3U108
Entrez Gene ID: (Human) 10865, (Mouse) 214855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support