Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
FIGURE: 1 / 2
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin alpha 1; Actin, alpha skeletal muscle; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Alpha-actin-1; Alpha-actin-2; alpha-cardiac actin; Cell growth-inhibiting gene 46 protein; nemaline myopathy type 3; smooth muscle alpha-actin
Gene Aliases: 0610041G09Rik; a-SMA; AA959943; AAT6; ACTA; Acta-2; ACTA1; ACTA2; Acts; ACTSA; Actsk-1; ACTVS; alphaSMA; ASMA; CFTD; CFTD1; CFTDM; GIG46; MPFD; MYMY5; NEM1; NEM2; NEM3; SHPM; SMalphaA
UniProt ID: (Human) P68133, (Human) P62736, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737
Entrez Gene ID: (Human) 58, (Human) 59, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support