Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200-1:1,000 | View 4 publications 4 publications |
Immunohistochemistry (IHC) |
- | View 1 publication 1 publication |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:500 | - |
Immunoprecipitation (IP) |
0.5-4.0 µg | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
BAMBI Fusion Protein Ag8974 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.37 mg/mL |
Storage conditions |
-20°C |
BAMBI (BMP and activin membrane-bound inhibitor homolog, NMA) is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP and activin membrane-bound inhibitor homolog; BMP and activin membrane-bound inhibitor, homolog; Kinase-deficient TGFbeta superfamily receptor subunit; Non-metastatic gene A protein; Putative transmembrane protein NMA
Gene Aliases: BAMBI; NMA
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support