Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SSVKKQKALV AKVSQREEMV KKCLGELTEV CKSLGKVFGV HYFNIFNTVT LKKLAESLSS DPEVLLQIDG VTEDKLEKYG AEVISVLQKY SEWTSPAEDS
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bloom syndrome protein; Bloom syndrome, RecQ helicase-like; DNA 3'-5' helicase BLM; DNA helicase, RecQ-like type 2; MGC126616; MGC131618; MGC131620; RecQ protein-like 3; RecQ-like DNA helicase BLM; RecQ2
Gene Aliases: BLM; BS; RECQ2; RECQL2; RECQL3
UniProt ID: (Human) P54132
Entrez Gene ID: (Human) 641
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support