Invitrogen
MA1-7004 detects Bacillus anthracis.
MA1-7004 has been successfully used in Western blot and sandwich ELISA procedures as a capture antibody, with MA1-7476 for detection.
The MA1-7004 immunogen is purified recombinant 90 kDa spore antigen.
Anthrax toxin is a three-protein exotoxin secreted by virulent strains of the bacterium Bacillus anthracis, the causative agent of anthrax. Anthrax toxin is composed of a cell-binding protein, known as protective antigen (PA), and two enzyme components, called edema factor (EF) and lethal factor (LF). These three protein components act together to impart their physiological effects. Assembled complexes containing the toxin components are endocytosed. In the endosome, the enzymatic components of the toxin translocate into the cytoplasm of a target cell. Once in the cytosol, the enzymatic components of the toxin disrupts various immune cell functions, namely cellular signaling and cell migration. The toxin may even induce cell lysis, as is observed for macrophage cells. Anthrax toxin allows the bacteria to evade the immune system, proliferate, and ultimately kill the host animal. Research on anthrax toxin also provides insight into the generation of macromolecular assemblies, and on protein translocation, pore formation, endocytosis, and other biochemical processes.