This antibody detects endogenous protein at a molecular weight of 22 kDa.
Purity is >95% by SDS-PAGE.
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C EBP; C/EBP gamma; CCAAT/enhancer binding protein (C/EBP), gamma; CCAAT/enhancer binding protein ,gamma; CCAAT/enhancer-binding protein gamma; CEBP gamma; G-CSF promoter element 1 binding protein; GPE1-binding protein; GPE1-BP; GPE1-BP (C/EBP gamma); Granulocyte colony-stimulating factor promoter element 1-binding protein; homolog to a human CCAAT/enhancer binding protein - gamma; IG/EBP-1; Immunoglobulin enhancer-binding protein 1
Gene Aliases: C/EBP; C/EBP[g]; C/EBPgamma; CEBPG; CEBPRNA; GPE1-BP; GPE1BP; Ig/EBP; IG/EBP-1
UniProt ID: (Human) P53567, (Rat) P26801, (Mouse) P53568
Entrez Gene ID: (Human) 1054, (Rat) 25301, (Mouse) 12611
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support