Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GOLGA7B gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein targeting to membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: golgi autoantigen, golgin subfamily a, 7B; golgin A7 family, member B; Golgin subfamily A member 7B
Gene Aliases: 4933417O08Rik; AI839934; bA451M19.3; bA459F3.4; C10orf132; C10orf133; GOLGA7B
UniProt ID: (Human) Q2TAP0, (Mouse) Q9D428
Entrez Gene ID: (Human) 401647, (Mouse) 71146, (Rat) 309378
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support