Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: SPVTLTKGTS AAHLNSMEVT TEDTSRTDVS EPATSGGAAD GVTSIAPTAV
Sequence homology: Human: 100%
The function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein DM4E3; Uncharacterized protein C11orf24
Gene Aliases: C11orf24; DM4E3; FP2568; UNQ1872/PRO4315
UniProt ID: (Human) Q96F05
Entrez Gene ID: (Human) 53838
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support