Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Putative methyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Probable methyltransferase-like protein 25
Gene Aliases: C12orf26; METTL25
UniProt ID: (Human) Q8N6Q8
Entrez Gene ID: (Human) 84190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support