Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYTITKGPS KLVAQRRTGP TQQQVEGRLG ELLKCRQPAP PTSQPPRAQP FAQPPGPWPL SSLAAGATAA GWWPSR (1-75 aa encoded by BC007346)
FAM195A gene ontology (GO) annotation includes protein binding and nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 195 member A; family with sequence similarity 195, member A; MAPK regulated co-repressor interacting protein 2; MAPK regulated corepressor interacting protein 2; Protein FAM195A
Gene Aliases: 9530058B02Rik; C16orf14; c349E10.1; FAM195A; MCRIP2
UniProt ID: (Mouse) Q9CQB2, (Human) Q9BUT9
Entrez Gene ID: (Mouse) 68241, (Human) 84331
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support