Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The exact function of C16orf48 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cp048; Enkurin domain-containing protein 1
Gene Aliases: AI606951; C16orf48; DAKV6410; E130303B06Rik; ENKD1; UNQ6410/PRO21183
UniProt ID: (Human) Q9H0I2, (Mouse) Q7TSV9
Entrez Gene ID: (Human) 84080, (Mouse) 102124
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support