Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MP11; Myelin protein of 11 kDa; noncompact myelin associated protein; Noncompact myelin-associated protein
Gene Aliases: A330049M08Rik; AA545175; C1orf130; MP11; NCMAP
UniProt ID: (Human) Q5T1S8, (Rat) F1M2Z5, (Mouse) Q99JS0
Entrez Gene ID: (Human) 400746, (Rat) 689826, (Mouse) 230822
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support