Search Thermo Fisher Scientific
PA5-16925 targets C3c Complement in IF applications and shows reactivity with Human samples.
The PA5-16925 immunogen is c3c complement from human serum.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.