Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MASGKDTCPT LPKLTNNCSD ESLYKSANKY EEIHLPRFSL KQGMIPRRYV MPWKENMIFR NVNLKQAEVC GIHTGPLEDS LFLNHSERLC HGEDRKVVFQ KGPPEIKIAD MPLHSPLSRY QSTVISHGFR RRLV
TEX43 is a protein coding gene that is enriched in the testis tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sperm-associated microtubule inner protein 10; testis specific expressed gene 7; Testis-expressed protein 43
Gene Aliases: C5orf48; SPMIP10; TEX43; Tseg7
UniProt ID: (Human) Q6ZNM6
Entrez Gene ID: (Human) 389320
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support