Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
C5AR2 is an orphan member that belongs to anaphylatoxin receptor family. It is a seven transmembrane G-protein-coupled receptor consisting of an N-terminal N-glycosylation site, several conserved cysteines and several potential C-terminal phosphorylation sites. It binds to C5a and C5a-des-Arg and serves as a highly regulated scavenger receptor with a negative modulatory influence on C5aR-mediated inflammation. Northern Blot analysis suggests it is co- expressed along with C5aR in different tissues and cells including neutrophils/macrophages, mast cells, dendritic cells, brain, lung, heart, kidney, liver, ovary and testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C5a anaphylatoxin chemotactic receptor 2; C5a anaphylatoxin chemotactic receptor C5L2; Complement component 5a receptor 2; G protein-coupled receptor 77; G protein-coupled receptor C5L2; G-protein coupled receptor 77
Gene Aliases: C5AR2; C5L2; GPF77; GPR77
UniProt ID: (Human) Q9P296
Entrez Gene ID: (Human) 27202
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support