Search Thermo Fisher Scientific
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 µL of deionized water.
CALHM3 is a pore-forming subunit of a voltage-gated ion channel.