Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Calreticulins, such as Calreticulin 3, are Ca(2+)-binding chaperones localized mainly in the endoplasmic/sarcoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calreticulin-2; Calreticulin-3; Calsperin; cancer/testis antigen 93; testis secretory sperm-binding protein Li 226n
Gene Aliases: CALR3; CMH19; CRT2; CT93
UniProt ID: (Human) Q96L12
Entrez Gene ID: (Human) 125972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support