Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RQRVQDLMA QMQGPYNFIQ DSMLDFENQT LDPAIVSAQP MNPTQNMDMP QLVCPPVHSE SRLAQPNQVP VQPEATQVPL VSSTSEGYTA SQPLYQPSHA TEQRPQKEPI DQIQATISLN TDQTTASSSL PAASQPQVFQ AGTSKPLHSS GINVNAAPFQ SMQTVFNMNA PVPPVNEPET LKQQNQYQAS YNQSFSSQPH QVEQTELQQE QLQTVVGTYH GSPDQSHQVT GNHQQPPQQN TGFPRSNQPY YNSRGVSRGG SRGARGLMNG YRGPANGFRG GYDGYRPSFS NTPNSGYTQS QFSAPRDYSG YQRDGYQQNF KRGSGQSGPR GAPRGRGGPP RPNRGMPQMN TQQVN (356-709 aa encoded by BC001731)
GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activation/proliferation-associated protein 1; caprin 1; Caprin-1; Cell cycle-associated protein 1; cytop; Cytoplasmic activation- and proliferation-associated protein 1; cytoplasmic activation/proliferation-associated protein 1; cytoplasmic activation/proliferation-associated protein-1; GPI-anchored membrane protein 1; GPI-anchored protein p137; GPI-p137; Membrane component chromosome 11 surface marker 1; membrane component, chromosome 11, surface marker 1; p137GPI; RNA granule protein 105
Gene Aliases: AL022980; Caprin-1; CAPRIN1; G5E5; GPI p137; Gpiap; GPIAP1; GPIP137; GRIP137; M11S1; Mmgpip137; P137; p137GPI; RNG105
UniProt ID: (Human) Q14444, (Mouse) Q60865, (Rat) Q5M9G3
Entrez Gene ID: (Human) 4076, (Mouse) 53872, (Rat) 362173
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support