Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SLTRQMEKDE TVSDCSPHIA NIGRLVEDME NKIRSTLNEI YFGKTKDIVN GLRSVQTFAD KSKQEALKND LVEALKRKQQ C
CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cap Z; capping protein (actin filament) muscle Z-line, beta; CapZ beta; F-actin-capping protein subunit beta; MGC104401; MGC129749; MGC129750; RP4-657E11.7
Gene Aliases: CAPB; CAPPB; CAPZ; CAPZB
UniProt ID: (Human) P47756
Entrez Gene ID: (Human) 832
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support